Search
EVOS cell imaging systems are designed to eliminate the complexities of microscopy without compromising on performance. These units combine all features of a digital inverted microscope workstation into a single, compact device that can be mastered in minutes. Whether you’re capturing tissue images for publication or counting cells in a culture hood, these systems are designed to be easier, smarter, and faster.
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
Thermo Fisher Scientific offers a transmitted-light imaging solution to fit your application—you choose the system that is right for you based on the imaging applications you want to run. Any or all of the following could influence your choice of platform:
Use the selection guide below to find a system that fits you.
![]() EVOS XL Core Perfect for cell culture and routine cell maintenance | ![]() EVOS XL Perfect for more advance colorimetric assays | ![]() EVOS FL Auto Perfect for a variety of advanced automated imaging applications | |
| High-resolution LCD display | 12.1” | 15” | 22” Touch screen monitor |
|---|---|---|---|
| Camera | Color CMOS | Color CMOS | Dual monochrome (CCD) and color (CMOS) |
| Transmitted-light (brightfield & phase-contrast) | ✓ | ✓ | ✓ |
| Epifluorescence imaging | ✓ | ||
| Fits in tissue culture hood | ✓ | ||
| Time-lapse imaging | ✓ | ✓ | |
| Onstage incubator | ✓ (optional) | ||
| Autofocus | ✓ | ||
| High-resolution image mapping | ✓ | ||
| Cell counting | ✓ | ✓ | |
| Weight | 9.1 kg (20.1 lb) | 15.3 kg (33.7 lb) | 20 kg (44.1 lb) |
| Dimensions | H: 55.3 cm (21.0") W: 31.8 cm (12.5") D: 40.6 cm (16.0") | H: 57.8 cm (22.8") W: 35.5 cm (14.0") D: 47.0 cm (18.5") | H: 32.2 cm (12.7") W: 34.3 cm (13.5") D: 47.2 cm (18.6") |
NewCell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow.