Streptomycin Sulfate
Streptomycin sulfate, an aminoglycoside used for bacterial selection, binds to the small ribosomal subunit and interferes with protein biosynthesis.
Application(s): Selection of resistant bacteria.
Application(s): Selection of resistant bacteria.