Ultra Low IgG Fetal Bovine Serum
Ultra low IgG fetal bovine serum has been chromatographically "purified" of IgG, an important factor in isolating organisms that may be inhibited by pre-existing levels of IgG or quantitative IgG immunoassays. The serum retains full biological activity.
  • Lowers Ig levels (< 5 μg/ml) in fetal bovine serum while preserving comparable performance to regular fetal bovine serum
  • BVD antibody titer is low or not detectable
  • Expert technical support
  • Now available in our innovatively designed, aliquot-free Gibco One Shot FBS 50 mL bottle
Gibco media & reagents