Premade Clones / Clone Collections
By making clone manuals available online and no longer shipping them with every order, we are significantly reducing paper usage. You can now access, review, and download needed clone information from one easy location, whenever you need it. Access clone manuals here.
- For the research application of each clone type, check out the Clone Collection Selection Guide.
Featured clone collections categories
Ultimate™ ORF clones
Full-insert sequenced open reading frames (ORFs) of human and mouse clones in a Gateway® entry vector. Ultimate™ ORF clones offer the highest utility for your downstream analysis needs in plates or tubes and with different QC levels to match your needs.
- Search for your gene of interest
- Learn more about Ultimate™ ORF clones
- Learn more about Ultimate™ ORF clones LITE
BAC/PAC clones
A repository of clones from genomic libraries which contain DNA inserts in excess of 100 to 300 kb. Sources for BAC clones include human and mouse tissues. Sources for PAC clones include human, mouse and rat tissues.
- Search for your BAC/PAC clone
- Learn more about BAC/PAC clones
Yeast deletion clones
A collection of clones where each yeast deletion represents a unique gene-knockout of the S. cerevisiae genome. The yeast deletion strains are available in the form of Mat-a haploid, Mat-alpha haploid, heterozygous diploid, and homozygous diploid.
- Search for your Yeast deletion clone
- Learn more about Yeast deletion clones
Yeast GFP Clones
A collection of S. cerevisiae GFP-tagged open reading frame clones.
- Search for your Yeast GFP clone
- Learn more about Yeast GFP clones
Support
Cloning Support Center
Find tips, troubleshooting help, and resources for your cloning applications
DISCLAIMER—Clone Collections:
These clone collections were generated by groups outside of Invitrogen. The quality of the collections is largely dependent upon what was received by these groups. Specific clone information and plate coordinates originated from the suppliers of these clone collections and have not been confirmed by Invitrogen.
Invitrogen has instituted quality procedures to ensure that clones are picked from the identified well in a plate, grown on the correct antibiotic, and do not have phage contamination. Due to the quality of the information provided to Invitrogen, the clone you receive might not match the expected clone. If this occurs, please contact Technical Support.
Please note the Ultimate ORF Collection was generated by Invitrogen and all data associated with these clones has been verified by Invitrogen prior to release for sale. Invitrogen fully guarantees each Ultimate ORF Clone to match the accession number associated with the clone ID.