COL16318-Staticdisplay-social-banners-Connected-Consumables-head-image
Real-time PCR (also known as quantitative PCR or qPCR) is the simultaneous amplification and detection/quantification of nucleic acids using the polymerase chain reaction (PCR). qPCR is a versatile technique and a powerful tool to support your research applications. Find optimized real-time PCR master mixes, reagents, and kits to power your experiments here.
Recommended qPCR master mixes by application
Chemistry
Application
Start with..
Applied Biosystems™ TaqMan™*
Gene
expression
TaqMan™ Fast Advanced Master Mix
for 2-step
TaqMan™ Fast Virus Master Mix  
for 1-step virus detection and gene expression
Applied Biosystems™ TaqPath™**
(Clinical-grade TaqMan)
Gene
expression
TaqPath™ qPCR Master Mix
for 2-step
TaqPath™ 1-step RT-qPCR Master Mix
for 1-step
TaqPath™ 1-step Multiplex Master Mix
for 1-step & multiplex
Genotyping
TaqPath™ ProAmp Master Mix
for human or animal samples only (for plant samples, please visit our Agrigenomics section)
Applied Biosystem™ SYBR™
Gene
expression
PowerUp SYBR™ Green Master Mix
Sample-to-answer
With Sample-to-Answer Kits, crude sample lysates can be used directly in real-time PCR or one-step RT-PCR. No nucleic acid purification is required:
Looking for qPCR reagents for molecular diagnostic test development? 
See our molecular diagnostic section
Looking for qPCR multiplexing solutions? 
See our qPCR multiplex applications section
Real-time PCR master mix selection tool
Select from our full selection of optimized Applied Biosystems™ and Invitrogen™ real-time PCR reagents to find the one that’s right for your desired application, detection chemistry, format, and running mode.
{{column.ids[1]}}
  • {{value}}
Application Detection chemistry Type/ format Multiplexing Mode/ speed Lab type Instruments Recommended
{{row["application+0"]}} {{row["chemistry+1"]}} {{row["type+2"]}} {{row["multiplexing"]}} {{row["mode"]}} {{row["lab"]}} {{row.instruments}} {{row.title}}
* For Research Use Only. 
** General purpose reagent. For laboratory use. (Can also be used for research.)