NovaBright Reporter Gene Assay Selection Guide
NovaBright Beta-gal System | NovaBright Dual Beta-Gal & Luciferase System | NovaBright Luciferase System | NovaBright SEAP System | |
---|---|---|---|---|
Reporter Enzyme(s) | Beta-galactosidase | Luciferase, beta-galactosidase | Luciferase | Secreted placental alkaline phosphatase |
Key Attributes | Homogeneous assay does not require removal of culture medium | Detection of luciferase and beta-galactosidase from the same cell extract | Ideal for high-throughput screening | No cell lysis required |
Substrate | Galacton-Star®, Gal-Star®, Galacton-Plus® | Luciferin, Galacton-Plus® | Luciferin | CSPD® |
Duration of Light Emission | 60–90 min | 60–90 min | 60–90 min | 1–2 hr |
Reagent Injection | Not required | Required | Not required | Not required |
Special Features | Designed for assays on cells cultured in luminometer plates for high-throughput screening. Choice of reaction buffers for lysis of mammalian or yeast and mammalian cells. | Two assays from one cell extract enables better precision for normalizing transfection efficiency. | Linear signal from 50 fg to 100 ng, stable light emission from 10 minutes to 4-5 hours. | Cells remain viable; useful for stable transfectants, time course studies, etc. |
See all Beta-gal Kits | See all Dual Systems | See all Luciferases | See all SEAP Kits |
Download more information
- Download the Chemiluminescent Reporter Gene Assays Brochure
For Research Use Only. Not for use in diagnostic procedures.