34090-X-Ray-Film-230
Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based Western blots and other chemiluminescent protein or nucleic acid assay methods.
CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.
Highlights
  • High performance – just as sensitive as other commercially available autoradiography films
  • Affordable – costs up to 75% less than other brands of comparable quality X-ray film
  • Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets
  • Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate
Comparison between Thermo Scientific CL-XPosure Film and Kodak Brand X-ray films.
Feature Thermo Scientific
CL-XPosure Film
Kodak X-Omat™
Blue (XB) Film
Kodak BioMax™
MR-1
Supplier
(Part No.)
Thermo Scientific
(34090)
Perkin Elmer
(NEF586001EA)
Perkin Elmer
(8941114001)
Color of Film Blue Blue Gray
Isotopes detected 35S, 32P, 14C All isotopes 35S, 32P, 14C
ECL Detection Yes Yes Yes
Notes High sensitivity & contrast, med. speed Requires longer exposure than other Kodak films Maximum resolution, clarity and sensitivity
Cost per Sheet Low Medium High
Conclusion Best Good Better
† Source: 2010 Online Catalogs
Product Details
Looking for replacements for Amersham™ Hyperfilm™ Products from GE Healthcare?
Discontinued Hyperfilm Product » Equivalent CL-XPosure Film
28-9888-22 Hyperfilm Blue
12.7 x 17.8cm (5 x 7in.)
100 sheets
» 34088 CL-XPosure Film
5 x 7in. (12.5 x 17.5cm)
50 sheets
» 34090 CL-XPosure Film
5 x 7in. (13 x 18cm)
100 sheets
28-9888-21 Hyperfilm Blue
18 x 24cm (7.1 x 9.5in.)
100 sheets
» 34089 CL-XPosure Film
7 x 9.5in. (18 x 24cm)
100 sheets
28-9888-20 Hyperfilm Blue
20.3 x 25.4cm (8 x 10in.)
100 sheets
» 34091 CL-XPosure Film
8 x 10in. (20 x 25cm)
100 sheets
28-9068-40 Hyperfilm ECL
24 x 30cm (10 x 12in.)
50 sheets
» 34097 CL-XPosure Film
9.5 x 11.8in. (24 x 30cm)
100 sheets
28-9068-41 Hyperfilm ECL
35 x 43cm (11.8 x 15.8in.)
50 sheets
» 34099 CL-XPosure Film
14 x 17in. (35 x 40cm)
100 sheets
28-9068-47 Hyperfilm MP
24 × 30cm (10 x 12in.)
Single film bag, 50 sheets
» 34097 CL-XPosure Film
9.5 x 11.8in. (24 x 30cm)
100 sheets
28-9068-48 Hyperfilm MP
35 × 43cm (11.8 x 15.8in.)
Single film bag, 50 sheets
» 34099 CL-XPosure Film
14 x 17in. (35 x 40cm)
100 sheets
Comparison between Thermo Scientific CL-XPosure Film and Kodak Brand X-ray films.
Feature Thermo Scientific
CL-XPosure Film
Kodak X-Omat™
Blue (XB) Film
Kodak BioMax™
MR-1
Supplier
(Part No.)
Thermo Scientific
(34090)
Perkin Elmer
(NEF586001EA)
Perkin Elmer
(8941114001)
Color of Film Blue Blue Gray
Isotopes detected 35S, 32P, 14C All isotopes 35S, 32P, 14C
ECL Detection Yes Yes Yes
Notes High sensitivity & contrast, med. speed Requires longer exposure than other Kodak films Maximum resolution, clarity and sensitivity
Cost per Sheet Low Medium High
Conclusion Best Good Better
† Source: 2010 Online Catalogs
Product Details
Looking for replacements for Amersham™ Hyperfilm™ Products from GE Healthcare?
Discontinued Hyperfilm Product » Equivalent CL-XPosure Film
28-9888-22 Hyperfilm Blue
12.7 x 17.8cm (5 x 7in.)
100 sheets
» 34088 CL-XPosure Film
5 x 7in. (12.5 x 17.5cm)
50 sheets
» 34090 CL-XPosure Film
5 x 7in. (13 x 18cm)
100 sheets
28-9888-21 Hyperfilm Blue
18 x 24cm (7.1 x 9.5in.)
100 sheets
» 34089 CL-XPosure Film
7 x 9.5in. (18 x 24cm)
100 sheets
28-9888-20 Hyperfilm Blue
20.3 x 25.4cm (8 x 10in.)
100 sheets
» 34091 CL-XPosure Film
8 x 10in. (20 x 25cm)
100 sheets
28-9068-40 Hyperfilm ECL
24 x 30cm (10 x 12in.)
50 sheets
» 34097 CL-XPosure Film
9.5 x 11.8in. (24 x 30cm)
100 sheets
28-9068-41 Hyperfilm ECL
35 x 43cm (11.8 x 15.8in.)
50 sheets
» 34099 CL-XPosure Film
14 x 17in. (35 x 40cm)
100 sheets
28-9068-47 Hyperfilm MP
24 × 30cm (10 x 12in.)
Single film bag, 50 sheets
» 34097 CL-XPosure Film
9.5 x 11.8in. (24 x 30cm)
100 sheets
28-9068-48 Hyperfilm MP
35 × 43cm (11.8 x 15.8in.)
Single film bag, 50 sheets
» 34099 CL-XPosure Film
14 x 17in. (35 x 40cm)
100 sheets
References
  1. De Ioamnes, P., et al. (2004). J. Biol. Chem. 279, 26134-26142.
  2. Perlman, A., et al. (2004). Ann. Clin. Lab. Sci. 34, 277-286.
  3. Tikhonov, I., et al. (2003). J. Virol. 77, 3157-3166.
Resources
Protein Electrophoresis and Western Blotting Support Center – Find tips, troubleshooting help, and resources for your western workflow.