Description
Ambion® In Vivo siRNA molecules are chemically modified, 21-mer, double-stranded siRNAs that are recognized by the RNA-induced silencing complex (RISC) to mediate inhibition of a target gene. Proprietary chemical modifications allow Ambion® In Vivo siRNAs to overcome many in vivo–specific obstacles, ensuring their effectiveness and stability in vivo. Ambion® In Vivo siRNAs are at least 100x more stable in 90% mouse serum than unmodified siRNAs.
Ambion® In Vivo siRNAs, the new standard for in vivo RNAi applications, offer:
  • High stability against nucleases
  • No induction of the interferon response
  • Easy tracking of administered siRNAs
  • Combined knockdown effectiveness and stability
Catalog # Purification Size
4457298 Standard Purity 5 nmol
4457302 HPLC purified 250 nmol
4457304 HPLC purified 1 umol
4457306 HPLC purified 10 umol
4457308 HPLC In Vivo Ready 250 nmol
4457310 HPLC In Vivo Ready 1 umol
4457312 HPLC In Vivo Ready 10 umol
Documents
Product Literature
Search More Than 200,000 Ambion® siRNAs
Meet the Inventor: Silencer® Select siRNA technology
Search more than 200,000 Ambion® siRNAs from Life Technologies
Watch the video and learn how >
Help Center
Order Support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical Support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests
Product Categories
Related Applications
Related Products
Description
Ambion® In Vivo siRNA molecules are chemically modified, 21-mer, double-stranded siRNAs that are recognized by the RNA-induced silencing complex (RISC) to mediate inhibition of a target gene. Proprietary chemical modifications allow Ambion® In Vivo siRNAs to overcome many in vivo–specific obstacles, ensuring their effectiveness and stability in vivo. Ambion® In Vivo siRNAs are at least 100x more stable in 90% mouse serum than unmodified siRNAs.
Ambion® In Vivo siRNAs, the new standard for in vivo RNAi applications, offer:
  • High stability against nucleases
  • No induction of the interferon response
  • Easy tracking of administered siRNAs
  • Combined knockdown effectiveness and stability
This product can be purchased by visiting AppliedBiosystems.com.
Direct ordering through the Life Technologies website will be available soon.
Documents
Product Literature
Search More Than 200,000 Ambion® siRNAs
Meet the Inventor: Silencer® Select siRNA technology
Search more than 200,000 Ambion® siRNAs from Life Technologies
Watch the video and learn how >
Help Center
Order Support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical Support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests