Simple data analysis and storage solutions
Torrent Suite software and hardware provide customizable, automated data analysis and storage solutions.
Request a quote
Torrent Suite Software and Torrent Server
Whether you are new to next-generation sequencing or an expert user, Torrent Suite Software makes it easy to get started sequencing on the Ion S5, Ion S5 XL, Ion PGM, and Ion Proton systems.
  • Easy-to-use software, preinstalled on the Torrent Server for automated sequencing data analysis, for the Ion S5, Ion S5 XL, Ion PGM, and Ion Proton systems
  • Intuitive, web-based interface through the Torrent Browser makes it fast and easy to plan, monitor, and view sequencing run results
  • Optimized for Ion Torrent raw data analysis, alignments, and variant calling using the Torrent Variant Caller plug-in
  • Expanded capabilities with Ion Torrent verified plug-ins available
  • Seamless integration with Ion Reporter Software, the Ion Reporter Server, and the Torrent Suite Storage System
1 Plan
pre-designed templates
Create a run plan and download directly to the sequencer.
Input your sequencing run information, including run details, sample IDs, and automated data analysis preferences to easily track and organize your data.
View screenshot
2 Monitor
Monitor sequencing
Monitor key sequencing metrics in real time
Watch your run progress in real time and monitor key metrics to help evaluate the status of the run.
View screenshot
3 Data
Run Sequence for RNA Sequencing
View run statistics and data analysis results.
View run results in a standard report format with easy-to-read graphics and key performance metrics.
View screenshot
4 Export
auto export data
Export data in industry standard formats.
Easily export your data to the Torrent Suite Storage System, Ion Reporter Software, or third party software using plug-ins.
View screenshot
Ion S5 Torrent Server specifications
Ion PGM Torrent Server specifications
Ion Proton Torrent Server specifications
For release notes, user guides and additional support, please visit our NGS Software Support page.
Torrent Storage NAS Device
High-throughput next-generation sequencing (NGS) can be fast and cost effective, but the rate of data generation often outgrows the original storage system. The Torrent Storage NAS Device, developed in collaboration with Dell, connects directly or via local area network to the Torrent Server and Ion Reporter Software for quick storage and retrieval of sequencing data and results.
  • Shared network storage—connect to any Ion Torrent sequencing platform and share data across multiple platforms via local area network
  • Fast and easy data retrieval—locally store and organize your sequencing data on-site
  • Dependable data storage—store valuable sequencing data with individual disk failure tolerance to help prevent data loss
View datasheet for more information.
Working with qPCR or Sanger sequencing instruments too? Check out Datasafe Solutions, the scalable, cross-platform compatible data storage solutions with advanced back-up and data recovery services
*On-site installation and configuration by a field service engineer and web-based training by a field bioinformatics specialist are included in the price to help ensure proper configuration.