Gene Expression by Sanger Sequencing
Sequencing and fragment analysis–based RNA assay methods give you the critical gene expression data you’re looking for using robust reagents and workflows.
![]() |
Serial Analysis of Gene Expression (SAGE™) Method used to compare expression of multiple genes in a given sample to create accurate relative expression profiles from whole-transcriptome analysis |
![]() |
High Coverage Gene Expression Profiling (HiCEP) Method for whole-transcriptome profiling, aimed at providing sensitive detection of a large proportion of transcripts for both known and unknown genes. |
Visistat reference component
For Research Use Only. Not for use in diagnostic procedures.