Search
We are dedicated to providing you with the latest insights and information on electron microscopy imaging and analysis for batteries. We offer a wide range of valuable resources, including on-demand webinar recordings, interviews with key industry leaders, and application documents. Whether you are a seasoned industry professional or just starting out in the field, this is the perfect place to access the latest information and stay up-to-date with the latest trends and technologies. From in-depth technical discussions to practical tips and advice, our resources are designed to help you navigate the complex world of battery research and production with ease. So, take some time to explore and discover all of the valuable resources we have to offer.
Challenge |
Technologies |
Solution |
Resources |
Avoid contamination of air-, moisture-, and/or beam-sensitive battery samples during preparation and sample transfer |
IGST workflow: DualBeam, SEM/Desktop SEM (in glove box), TEM, Avizo, CleanConnect |
Complete workflow to enable sample characterization of sensitive battery materials in their native state without contamination |
|
Detection of lithium is difficult using SEM, EDS, and TEM |
TOF-SIMS |
Accurately detect and map lithium in battery samples in 2D and 3D down to 10 ppm |
App note: Ion spectroscopy using TOF-SIMS on a Thermo Scientific Helios DualBeam |
TEM |
iDPC technology can clearly image light elements like lithium at atomic scale |
App note: Integrated Differential Phase Contrast on Talos S/TEM |
|
Characterize battery structure at different scales beyond the capacity of a single instrument |
CT, SEM, Raman, DualBeam, Avizo, EDS |
Correlative workflow allowing multiscale imaging and analysis of battery microstructure |
App note: Multiscale image-based control and characterization of lithium-ion batteries |
| App note: Multiscale 3D imaging solutions for Li-ion batteries | |||
Prepare a large 2D area on the sample surface with high polishing quality for 2D imaging and characterization |
DualBeam (Plasma FIB-SEM), EDS |
High-throughput automated spin mill with high surface quality |
App note: Large area automated sample preparation for batteries |
SEM, CleanMill |
CleanMill offers a dedicated workflow for air-sensitive samples, an ultra-high energy ion gun for fast polishing, and a cryogenic function to protect sample integrity |
||
Characterize key microstructure properties (like tortuosity) for electrode structure performance correlations |
DualBeam, EDS, TOF-SIMS, Avizo |
3D characterization of battery structure · Hardware to image 3D battery structure at different scales · Software to automate 3D imaging data collection · Thermo Scientific Avizo Software workflow for image analysis and quantification |
Blog post/video: Advancing lithium-ion battery technology with 3D imaging |
App note: Multiscale image-based control and characterization of lithium-ion batteries |
|||
Characterize beam-sensitive materials like SEI at nanoscale |
TEM, EDS, Avizo |
Nano- and atomic-scale characterization of energy materials · Cryo-EM workflow for accurate data collection with superior EDS performance · Avizo Software for structure quantification and visualization |
Brochure: Analytical solutions for battery and energy storage technology |
In situ kinetic analysis (like heating) via electron microscope |
SEM |
Multiple in situ heating stage choices with integrated software for Thermo Scientific SEMs to understand cathode synthesis mechanisms |
Brochure: Scanning electron microscopy for lithium battery research |
Characterize beam-sensitive separator samples without damage |
SEM/SDB |
Superior low-KeV imaging and a cryo-FIB milling solution allow characterization of separator microstructure in 2D and 3D |
App note: Strategies for accurate imaging on battery separator structure |
Probe intrinsic SEI within a coin cell via electron microscopy |
Laser Plasma FIB |
High-energy, high-milling rate laser enables direct cross-section milling to understand Li-metal cell degradation mechanism |
|
Understand stoichiometry of solid electrolyte film as a function of depth |
XPS |
XPS depth profiling can quantify elements at each depth |
Webinar: Understanding surface reactions of the solid electrolyte interface via advanced characterization techniques |
Measure electrode surface chemistry |
XPS |
XPS can quantify the chemical states present at the electrode surface |
App note: Analysis of electrode materials for lithium ion batteries |
Track the evolution of the SEI layer |
XPS |
Materials can be depth profiled using XPS and a cluster ion source to follow the development of the SEI layer after cycling |
|
In situ electrode cycling |
XPS |
Electrodes can be operated in situ to monitor spectral changes as they are charged and discharged |
|
Profile battery components ex situ without missing point-to-point variability across an area |
Raman |
Raman microscopy can be used to look at changes to materials and distributions of components that occurred during use or testing |
|
Identify phases and determine structures in anodes and cathodes |
Raman |
Raman microscopy can visually show the spatial distribution of different phases of the same material with different performance characteristics |
App note: Raman analysis of lithium-ion batteries – Part I: Cathodes |
App note: Raman analysis of lithium-ion batteries – Part II: Anodes |
|||
XRD |
XRD can help to identify and quantify specific polymorphic structures of interest to increase yield and efficiency |
Brochure: ARL EQUINOX 100 X-ray Diffractometers | |
Trace and map anode composition across charge and discharge cycles |
Raman |
Raman microscopy can be used for in situ monitoring of changes on electrode surfaces during charge/discharge cycles |
|
Confirm the presence of specific carbon allotropes as anode components and in hybrid materials |
Raman |
Raman spectroscopy is particularly adept at the analysis of allotropes of carbon, including carbon in hybrid materials |
App note: Raman analysis of lithium-ion batteries – Part II: Anodes |
Understand the association of ionic species and distribution of components in solid polymer electrolytes (SPEs) |
Raman |
Raman microscopy can be used to visualize the spatial distribution of components in SPEs and indicate ionic associations |
App note: Raman analysis of lithium-ion batteries – Part III: Electrolytes |
Study crystallinity, stability, and reactivity in battery materials |
XRD |
X-ray diffraction can determine the percentage of crystallinity vs amorphous content of the active material, as well as structural stability and repeatability in real time |
|
Follow charge/discharge reactions in situ |
XRD |
During charge/discharge, the cathode and anode of every battery cell undergo changes. XRD allows you to follow the changing phase composition and the evolution of the crystalline structure |
Abbreviations: Avizo = Avizo Software; CT = Computed tomography; DualBeam = Focused ion beam scanning electron microscopy (FIB-SEM); EDS = Energy-dispersive X-ray spectroscopy; FIB = Focused ion beam; FTIR = Fourier transform infrared spectroscopy; iDPC = Integrated differential phase contrast; IGST = Inert sample gas transfer; SDB = Small DualBeam; SEI = Solid electrolyte interface; SEM = Scanning electron microscopy; SPE = Solid polymer electrolytes; TEM = Transmission electron microscopy; TOF-SIMS = Time of flight secondary ion mass spectrometry; XPS = X-ray photoelectron spectroscopy; XRD = X-ray diffraction.
For Research Use Only. Not for use in diagnostic procedures.
Cannot serve request to /content/lifetech/north-america/en-us/home/materials-science/battery-research/resources.html on this server