Element Crystal hex
Curium • Actinide Rare Earth
Symbol: Cm
Date of discovery: 1944
Name origin: Pierre and Marie Curie
Appearance: unknown
Discoverer: G.T. Seaborg
Obtained from: man-made
Melting point: 1613 K
Boiling point: 3383 K
Density[kg/m3]: 13510
Molar volume: 18.05 × 10-6 m3/mol
Protons/Electrons: 96
Neutrons: 151
Shell structure: 2,8,18,32,25,9,2
Electron configuration: [Rn]5f76d17s2
Oxidation state: 3
Crystal structure: hexagonal
Although second in the series of the transuranium elements, curium was actually the third element in the series to be discovered. The element was first synthesized in 1944 and made in its elemental form for the first time in 1951. There are very few commercial applications for curium, but it may be useful in radioisotope thermoelectric generators in the future. The isotope curium-242 can generate about 2 watts of thermal energy per gram and is used in pacemakers, remote navigational buoys, and in space missions. If curium enters the body, it can be very destructive because it accumulates in bone tissue, which then destroys bone marrow and stops red cell formation. The element is named after the spouses Marie and Pierre Currie, who are recognized for discovering radium.
Curium • Actinide Rare Earth
Products
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.