Thulium • Lanthanide Rare Earth
Primary XPS region: Tm4d
Overlapping regions: N/A
Binding energies of common chemical states: N/A
Experimental information
N/A
Interpretation of XPS spectra
N/A
About this element
Element Crystal hex
Symbol: Tm
Date of discovery: 1879
Name origin: Thule, Scandinavia
Appearance: silvery white
Discoverer: Per Teodor Cleve
Obtained from: monazite
Melting point: 1545 K
Boiling point: 1950 K
Density[kg/m3]: 9.32
Molar volume: 19.1 × 10-6 m3/mol
Protons/Electrons: 69
Neutrons: 100
Shell structure: 2,8,18,31,8,2
Electron configuration: [Xe]4f136s2
Oxidation state: 3
Crystal structure: hexagonal
Thulium is a rare earth metal never found as a free element. It is found in small quantities with other rare earths in minerals, such as monazite. In its pure form, this metal has a bright, silvery lustre. Thulium is reasonably stable in air, but must be protected from moisture. The element is silvery-grey, soft, malleable, ductile, and can be cut with a knife. It currently has no commercial applications, although one of its isotopes, thulium-169, could be used as a radiation source for portable X-ray machines. First identified in Sweden by chemist Per Theodor Cleve, Thulium gets its name from the ancient name for Scandinavia, Thule.
element_thulium
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.