Product End-of-Use Recycling Solutions
Find recycling programs and responsible end-of-use options for your products. Circularity is a transformative approach to sustainability, where products are designed, used, and recycled continuously, minimizing waste and conserving resources. Our guide helps you manage the end-of-use phase of your products by providing information on various plastics and materials, along with information on where and who can recycle them. We make recycling accessible by helping you locate programs and major recyclers in your area.
Our aim is to provide you with the knowledge and resources needed to help support getting your products recycled and repurposed effectively. Our take-back programs reclaim instruments and components, avoiding wasteful disposal and meeting global recycling requirements. Let us help you recycle, preventing excess waste from occupying space in your lab or landfill.
Our guidance covers the recycling of plastic labware and instruments deemed nonhazardous according to local and country-specific regulations. The listed recyclers have a broad regional presence, helping to ensure accessibility for many users. Detailed instructions and further information on recycling can be obtained from the respective recycling programs and providers. This guidance currently applies to the United States and selected European countries only.*
Together, we can close the loop and build a more sustainable future.
Recycling Guide
Choose your product type to view recycling options. Key to resin types are also provided.
Labware and Supplies
Material type Product example Suggested recycling (location)
#2 HDPE
Nalgene Graduated HDPE Plastic Beakers
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#5 PP / PPCO
Nalgene Griffin Low-Form Plastic Beaker, Nalgene Plastic Graduated Cylinder Variety Pack 
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
# PS
Fisherbrand Lids for Fisherbrand Polystyrene Beakers with Pouring Lip
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
#7 PFA
Nalgene PFA Graduated Plastic Beakers with Handles,  Nalgene Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Not commonly available
#7 PMP
Nalgene PMP Griffin Low-Form Plastic Beakers, Nalgene PMP Economy Plastic Graduated Cylinders
Not commonly available
#7 PTFE
PTFE Beaker
Not commonly available
Material type Product example Suggested recycling (location)
Multilayer film
Bioprocess containers (BPCs), Bioprocess bags
Thermo Fisher BPC Recycling
(Canada, United States)
Silicone
Tubing
Not commonly available
TPE
Tubing
Not commonly available
Material type Product example Suggested recycling (location)
#1 PET
Nalgene Square PET Media Bottles, Gibco Bottles
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
#2 HDPE
High Density Polyethylene (HDPE) Bottles, Nalgene Tamper Evident Closure, Lids, Gibco Bottle Caps
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
 Veolia (Belgium, France, Germany, Ireland)
#4 LDPE
Low Density Polyethylene (LDPE) Bottles
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
#5 PP / PPCO
Polypropylene (PPCO) Bottles, Nalgene Polypropylene Pour Spout, Lids
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#7 PC
Polycarbonate (PC) Bottles
Polycarbin Services through Fisher Scientific  (CanadaUnited States)
#7 PETG
Nalgene Square PETG Media Bottles
Polycarbin Services through Fisher Scientific  (CanadaUnited States)
#7 FEP/PFA
Nalgene fluoropolymer FEP and PFA bottles
Not commonly available
Glass Type 1 (borosilicate)
Polycarbin Services through Fisher Scientific  (United States)
Glass Type 3 (soda-lime)
Septa bottles and jars,
Polycarbin Services through Fisher Scientific  (United States)
Material type Product example Suggested recycling (location)
#2 HDPE
Nalgene HDPE funnels, utility funnels
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#4 LDPE
Nalgene LDPE funnels
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Germany)
#5 PP
Nalgene PP analytical funnels, Nalgene Büchner Two-Piece PP funnels
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#5 PP with PES membrane
Syringe filters
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#6 PS with PES membrane
Disposable filter unit, bottletop filters
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Material type Product example Suggested recycling (location)
#2 HPDE
HDPE Carboy
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#4 LDPE
Nalgene LDPE Jugs with Closure
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
#5 PP / PPCO
Nalgene PPCO Jugs with Closures, Nalgene Wide-Mouth Straight Sided PPCO Jars with Closure
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#7 PC
Nalgene Straight-Sided Wide-Mouth Polycarbonate Jars with Closure
Polycarbin Services through Fisher Scientific  (Canada, United States)
Glass Type 1 (borosilicate)
Polycarbin Services through Fisher Scientific  (United States)
Glass Type 3 (soda-lime)
I-Chem amber glass jars, Boston round amber glass jugs
Polycarbin Services through Fisher Scientific  (United States)
Material type Product example Suggested recycling (location)
#5 PP / PPCO
Flasks, PCR Sustain Series plates
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#6 PS
Petri dishes, dishes, flasks,  Multidishes, Microwell 96-Well Microplate
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
Material type Product example Suggested recycling (location)
# 4 LDPE
Transfer Pipettes
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
#5 PP
Pipette tips
Thermo Scientific TipCycle Program
(United States)
Polycarbin Services through Fisher Scientific
(Canada, United States)
#5 PP
Pipette tip racks and boxes
Thermo Scientific TipCycle Program
(United States)
Thermo Scientific Cyclebox Program 
(United Kingdom)
Polycarbin Services through Fisher Scientific
(Canada, United Kingdom, United States)
Veolia
 (Germany)
#6 PS
Serological pipettes
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Material type Product example Suggested recycling (location)
#5 PP / PPCO
Microplates, deep-well plates, plate covers, KingFisher plastics, PCR Sustain Series plates
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#6 PS
Clear microplate lids, plates
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (France, Germany)
Material type Product example Suggested recycling (location)
Rigid PE
Transport container lids
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#5 PP / PPCO
Flip top vials, centrifuge tubes, microcentrifuge tubes, conical tubes, cryogenic tubes, screw cap storage tube, PCR Sustain Series tubes
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
Glass Type 1 (borosilicate)
Amber screw thread vials, I-Chem clear VOA glass vials
Polycarbin Services through Fisher Scientific  (United States)
Lab Equipment, Instruments and Accessories
Material type Product example Suggested recycling (location)
Mixed materials with copper
iBlot Transfer Stacks
Recyclers with E-Stewards or R2 certified
Mixed material electronic chips
Ion Torrent sequencing chips
See below
(Canada, European Union, United States)
Ion Torrent sequencing chips
The Ion Chip recycling program provides customers with a convenient way to recycle used Ion Torrent chips. To recover valuable metals from these chips, we’re partnering with two reputable, certified recyclers in the United States, Canada, and Europe to collect and process the chips. To recycle your Ion chips, simply mail your used chips to the location closest to you using the addresses below.
United States:
Gannon & Scott
Attn: Thermo Fisher Ion Torrent Chip Recycling
2113 East Sky Harbor Circle South,
Phoenix, AZ 85034
Tel: 800.556.7296
Gannon & Scott
Attn: Thermo Fisher Ion Torrent Chip Recycling
33 Kenney Drive
Cranston, RI 02920
European Union:
REUZEit BV
Coenecoop 630
2741PV, Waddinxveen
Zuid-Holland (ZH), Netherlands
Canada:
ERS International
Attn: Thermo Fisher Ion Torrent Chip Recycling
2450 Lawrence Ave East, UNIT 3-15
Toronto, Ontario
M1P2R7
Material type Product example Suggested recycling (location)
Mixed material reagent cartridge
SeqStudio Cartridge
Recyclers with E-Stewards or R2 certified
Material type Product example Suggested recycling (location)
Chromatography and mass spectrometry without coolants, refrigerants, or gases
Chromatography columns and systems, mass spectrometers
Recyclers with E-Stewards or R2 certified or WEEELABEX
High pressure liquid chromatography (HPLC) and ultra-high pressure liquid chromatography (UHPLC) instruments
Dionex Summit HPLC Systems, Thermo Scientific Accla LC Systems, Thermo Finnigan Surveyor HPLC Systems
Recyclers with E-Stewards or R2 certified or WEEELABEX
Mass Spectrometry
Triple Quadrupole Mass Spectrometer, HRAM Orbitrap Mass Spectrometer
Recyclers with E-Stewards or R2 certified
Trade in and up Mass Spectrometer program
Stainless Steel Columns
LC Columns in Chromatography Consumables
Triumvirate Environmental (Canada, United States)
Veolia (Canada, France, Germany, United Kingdom, United States)
Ion Chromatographyy Systems
Thermo Scientific Dionex,
ICS-5000, ICS-1500, ICS-1100 IC Systems
RecycleReturn IC Instrument Trade up (Canada, United States)
PEEK    Not commonly available
Material type Product example Suggested recycling (location)
Lab equipment with coolants or refrigerants
Refrigerators, incubators
Recyclers with E-StewardsR2 certified, or WEEELABEX
Lab equipment with coolants or refrigerants
Freezers
Recyclers with E-StewardsR2 certified, or WEEELABEX
ULT Trade up Program (United States) (contact sales rep.)
Material type Product example Suggested recycling (location)
Lab instruments without coolant refrigerants, or gases
Centrifuges, shakers, pipette, dispensers, evaporators
Recyclers with E-StewardsR2 certified, or WEEELABEX
Biological safety cabinets Biological safety cabinets BSC Decommission and Recycling Services (France, Germany, United Kingdom, United States, other countries upon request)
Centrifuges
Centrifuges
Recyclers with E-StewardsR2 certified, or WEEELABEX
Centrifuge Recycling Program (United States only)
Material type Product example Suggested recycling (location)
Lab instruments without coolant refrigerants, or gasses
Microscopes, DNA sequencing systems, laboratory automation
Recyclers with E-StewardsR2 certified, or WEEELABEX
Electron Microscopy
Electron Microscopes
Certified used electron microscope buyback, resale, and recycling program
Material type Product example Suggested recycling (location)
Systems without coolant refrigerants, or gases
qPCR, PCR systems
Recyclers with E-StewardsR2 certified, or WEEELABEX
Sample Prep
KingFisher Flex, KingFisher Presto or MagMax Express 96 Particle Processor
Recyclers with E-StewardsR2 certified, or WEEELABEX
KingFisher Trade-In Trade-up Program
PCR Instruments
Real-Time PCR, Digital PCR, QuantStudio PCR Systems
Recyclers with E-StewardsR2 certified, or WEEELABEX
Real-Time PCR and Digital PCR Instrument Trade-In Program
Refurbished qPCR Research Instruments
Thermal cyclers
Applied Biosystems Thermal Cyclers, ProFlex PCR Systems, Veriti Thermal Cyclers
Recyclers with E-Stewards or R2 certified
Trade-in / donate Thermal Cyclers
#5 PP / PPCO
Accessory plates for analytic systems, KingFisher plastic plates, PCR plates, PCR Sustain Series plates and tubes
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
Safety Products
Material type Product example Suggested recycling (location)
Nitrile Gloves
All glove brands
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Safety Clothing
Garments, Bouffants and hair nets, boot and shoe covers
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
#7 Polycarbonate
Reusable safety Glasses
Polycarbin Services through Fisher Scientific  (Canada, United States)
Packaging
Material type Product example Suggested recycling (location)
#1 PET
Clam shell packaging
Polycarbin Services through Fisher Scientific (Canada, United Kingdom, United States)
#4 LDPE Film
Plastic bags, shipping bubbles,
Polycarbin Services through Fisher Scientific (Canada, United Kingdom, United States)
Veolia (Germany)
#4 LDPE Flexible pouch with viscous gel refrigerants
Gel ice packs
Not commonly available
#6 PS
Clam shell packaging
Polycarbin Services through Fisher Scientific (Canada, United Kingdom, United States)
#6 EPS
Packing peanuts, EPS foam inserts (white or black foam)
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Cardboard corrugated
Packaging dividers
Readily recyclable
EPE
Flexible foam, spongy foam
Not commonly available
Triumvirate Environmental (case by case United States)
EPU
Rigid foam
Not commonly available
Multilayer film
Product bags
Not commonly available
Triumvirate Environmental (case by case United States)
Paper
Fill paper, product inserts,
Readily recyclable
Material type Product example Suggested recycling (location)
#2 HDPE
Plastic buckets and drums
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Belgium, France, Germany, Ireland)
#3 PVC
No longer used by Thermo Fisher for packaging
Not commonly available
#4 LDPE Film
Stretch wrap, stretch film
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Veolia (Germany)
#5 PP
Plastic banding, labels, packaging tape
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States)
Triumvirate Environmental (Canada, United States)
Veolia (Canada, France, Germany, United Kingdom, United States)
#6 EPS
Styrofoam coolers
Polycarbin Services through Fisher Scientific  (Canada, United Kingdom, United States
Cardboard corrugated
External shipping box, Paper Cooler, Gaylord pallet boxes
Readily recyclable
Cardboard corrugated with EPU foam inside
Shippers including E327s and E90s
Not commonly available
Fiber with metal closure
Fiber drums
Triumvirate Environmental (Canada, United States)
Veolia (Canada, France, Germany, United Kingdom, United States)
Paper
Shipping labels, inner boxes, shipping paperwork, inserts
Readily recyclable
Steel
Steel can, steel pails, 55-gallon metal drums
Triumvirate Environmental (Canada, United States)
Veolia (Canada, France, Germany, United Kingdom, United States)
Wood
Pallets, crates
Veolia (France, Germany, United Kingdom, United States)
Key to acronyms, resin identification codes and common names for plastics types
EPS (#6): Expandable polystyrene
FEP (#7): Fluorinated ethylene propylene 
HDPE (#2): High density polyethylene
LDPE (#1): Low density polyethylene
PC (#7): Polycarbonate
PE: Polyethylene
PES: Polyether Sulfone
PET/PETE (#1): Polyethylene terephthalate
PETG (#7): Polyethylene terephthalate glycol
PFA (#7): Perfluoroalkoxy
PMP (#7): Polymethylpentene
PTFE (#7): Polytetrafluoroethylene
PP (#5): Polypropylene
PPCO (#5): Polypropylene Copolymer 
PS (#6): Polystyrene
TPE: Thermoplastic elastomers
EPE:  Expanded polyethylene
EPU:  Expanded Polyurethane
Compliance with the European Union’s Waste Electrical & Electronic Equipment (WEEE) Directive
Products with this symbol are required to comply with the European Union’s Waste Electrical & Electronic Equipment (WEEE) Directive 2012/19/EU.
Thermo Fisher Scientific has various trade-in, returns, and recycling programs for instruments. 
For any inquiries, please reach out to your Thermo Fisher representative.
*Thermo Fisher Scientific and its affiliates make no assertions or guarantees as to the services/products that third-party recyclers offer or their accuracy with regard to processes, results or outcomes. Thermo Fisher Scientific takes no legal or financial responsibility for services, products or the statements made by third-party recyclers, either on this page or in its marketing materials. Each customer of these recyclers must make its own determinations in whether to utilize these entities for their intended purposes.
For Research Use Only. Not for use in diagnostic procedures.