Organism TypeHumanHuman lung fibroblast. These are diploid cell lines derived from embryonic normal human lung. ATCC # CCL-75
TissuesLung
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,Transient Transfection