Search
Jurkat (human T-cell leukemia) cells were treated with 10 µM EdU for 2 hours, stained with CD4-PE Cy7 (Cat. No MHCD0412), and detected according to the recommended staining protocol. The figures show a clear separation of proliferating cells which have incorporated EdU and non-proliferating cells which have not. Panel A shows data from cells labeled with Alexa Fluor™ 350 picolyl azide analyzed on a BD™ LSRII flow cytometer using UV excitation and a 450/50 nm bandpass emission filter; Panel B shows the same cells using 488 nm excitation and a 780/60 nm bandpass emission filter for detection of the CD45-Pacific Orange™ and UV excitation and a 450/50 nm bandpass emission fileter for detection of the Alexa Fluor 350 picolyl azide; Panel C shows the dual parameter plot of the Click-iT™ Plus EdU Alexa Fluor™ 350 and FxCycle™ Far Red. Data were collected and analyzed using a BD LSRII flow cytometer using UV excitation and a 450/50 nm bandpass emission filter for detection of the Alexa Fluor 350 picolyl azide and 633 nm excitation and a 660/20 nm bandpass emission filter for detection of the FxCycle Far Red fluorescence. This figure combines DNA content with EdU; cells that are positive for both labels are in S-phase of the cell cycle.