Search
전체검색
Performance of the Qubit® dsDNA HS Assay. The Qubit® dsDNA HS Assay has a linear detection range of 0.2–100 ng and is selective for dsDNA, even in the presence of an equal mass of RNA.
Increase in accuracy and precision using the Qubit® Go ›
Low protein-to-protein variation in the Qubit® protein assay Go ›