Column Compartment
Method transfer kit
The Vanquish Core HPLC system includes an integrated, tunable gradient delay volume from 0 – 230 µL. With the optional method transfer kit, the system can adjust the gradient delay volume up to 430 µL using easy software commands, simplifying your method transfer without altering the qualification status of your instrument.