We regret to inform you that the DynaChip Antibody Analysis System has been discontinued and permanently removed from the market. Please pursue contingency procedures as Invitrogen will not recommence distribution of this product line. Please contact your HLA Technical sales specialist with any further questions.
Automate your lab for efficient HLA antibody screening and analysis
Invitrogen’s DynaChip Systems provide fully integrated instrumentation, reagents and software for automated HLA antibody screening and analysis. Class I and Class II antibodies are analyzed simultaneously in one test. The protein chip format offers the familiarity of  an ELISA method while allowing for increased multiplexing abilities, improving the efficiency of your lab processes and results.