QuantStudio 5 Real-Time PCR System
Take charge of how you create and manage your real-time PCR resultsThe QuantStudio™ 5 Real-Time PCR System is ideal for all levels of users who need a high-performance instrument with features for maximum experiment control. With built-in security features, locked workflows, user account management and options for Web browser-based or desktop analysis, the QuantStudio 5 Real-Time PCR System helps connect you to your data more easily than ever before and offers uncompromising performance and quality.
|
||
The QuantStudio 5 Real-Time PCR System is available with 96-well thermal blocks with either 0.1 or 0.2 mL well volume or a 384-well thermal block.
Get started quickly—The factory-calibrated system is delivered in a user-friendly package, enabling quick installation and immediate use.
Enable standard operating procedures and compliance with ease—User account management, locked protocol templates, in-run quality control (QC) feedback, QC traceability of consumables, and software pause features offer control of experimental data. Real-Time Data Mark-Up Language (RDML) export is available for compatibility with MIQE guidelines.
Save time with efficient experiment runs—Applied Biosystems™ VeriFlex™ technology offers 6 independent temperature zones for PCR optimization, and fast thermal cycling enables you to get results in as little as 30 minutes.
Maximum multiplexing you can trust—Applied Biosystems™ OptiFlex™ technology with 6 decoupled channels with 21 filter combinations and white LED enables optimal well-to-well and instrument-to-instrument accuracy and maximum multiplexing on the QuantStudio 5 96-well block system.
Get connected to your data more easily and quickly than ever before, with a real-time PCR system that offers an interactive touch screen for a modern usability experience similar to mobile devices and the option to choose Web browser–based software enabled through the Thermo Fisher Cloud. With the cloud, you can:
- Remotely monitor your runs and access data anytime, anywhere*
- Analyze sophisticated data sets quickly
- Store results in a secure data storage space
- Share and collaborate with colleagues at your institution or around the world
For peace of mind, the Thermo Fisher Cloud offers high industry-standard security, so your work remains secure. We have partnered with Amazon Web Services® (AWS) to offer a robust platform.
*Where the Web is accessible
| Repair Center service plans | On-site service plans | ||||
|---|---|---|---|---|---|
| Service plans at a glance | Applied Biosystems® Repair Center Support Plus | Applied Biosystems® Repair Center Support Plus Care | Applied Biosystems® Maintenance Plus | Applied Biosystems® Assurance | Applied Biosystems® Complete |
| On-site response time | Target 2 business days* | Guaranteed 2 business days* | Guaranteed next business day* | ||
| Scheduled on-site planned maintenance (PM) | Yes | Yes | Yes | Yes | |
| Remote diagnostics | Yes | Yes | Yes | Yes | Yes |
| Parts, labor, and travel for repair included | Yes | Yes | Yes | Yes | Yes |
| Computer repair and replacement included | Yes | Yes | 10% discount optional add-on in selected regions | Yes | Yes |
| Priority access to Tech Support Mon–Fri, 8 a.m.—5 p.m. local time |
Yes | Yes | Yes | Yes | |
| Priority access to Remote Service Engineer | Yes | Yes | Yes | Yes | |
| Requalification post-PM and critical repairs | Yes | ||||
| Field Application Scientist consultation | Yes | ||||
| Loaner instrument issued during repair (Repair Center plans only) | Yes | Yes | |||
*Instrument Management not available in all regions.
QuantStudio Family of Real-Time PCR and Digital PCR Systems
Product brochure: QuantStudio 3 and QuantStudio 5 Real-Time PCR Systems
Your Innovative Research: QuantStudio 5 Real-Time PCR System
Application Note: QuantStudio 3 and 5 Real-Time PCR Systems
Your Innovative Research: Expression of oncogenic microRNAs
Application Note: Protein Thermal Shift Technology
For Research Use Only. Not for use in diagnostic procedures.