Technical Resources - Media Formulations
Human Plasma-Like Medium (HPLM)
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0722.5210.3
L-Alanine89.0938.3090.43000337
L-Arginine HCl210.6623.1730.1100019
L-Asparagine Anhydrous132.126.6060.05
L-Aspartic acid133.12.6620.02
L-Cysteine HCl H2O175.637.0250.03999886
L-Cystine240.324.030.1
L-Glutamic Acid147.1311.770.07999728
L-Glutamine146.1580.3830.55000347
L-Histidine HCl H2O209.6323.0590.10999857
L-Isoleucine131.179.1820.07000077
L-Leucine131.1720.9870.15999848
L-Lysine HCl182.6536.530.2
L-Methionine149.214.4760.029997988
L-Phenylalanine165.1913.2150.07999879
L-Proline115.1323.0260.2
L-Serine105.0915.7640.15000476
L-Threonine119.1216.6770.14000167
L-Tryptophan204.2312.2540.06000098
L-Tyrosine F.B.181.1914.4950.079998896
L-Valine117.1525.7730.22
Vitamins
Biotin244.310.28.186321E-4
Choline chloride139.623.00.021486893
D-Calcium pantothenate476.540.255.2461494E-4
Folic Acid441.41.00.0022655188
Niacinamide122.131.00.008187996
Para-Aminobenzoic Acid137.141.00.0072918185
Pyridoxine HCl205.641.00.0048628673
Riboflavin376.370.25.31392E-4
Thiamine HCl337.271.00.0029649837
Vitamin B121355.380.0053.6890021E-6
i-Inositol180.1635.00.19427176
Inorganic Salts
Ammonium Chloride53.492.140.04000748
Calcium Chloride Anhyd110.98260.8032.35
Calcium Nitrate 4H2O236.159.4460.040000003
Magnesium Chloride (anhydrous)95.2145.7010.4800021
Magnesium Sulfate Anhydrous120.3642.130.35003325
Potassium Chloride74.55307.6554.1268272
Potassium Phosphate Monobasic136.092.00.014696158
Sodium Bicarbonate84.012016.2424.0
Sodium Chloride58.446216.2106.368935
Sodium Phosphate Dibasic Anhydrous141.96135.0050.9510073
Reducing Agents
Glutathione Reduced307.337.6830.024999188
Other Components
Acetone58.083.4850.06000344
Creatine149.155.2450.03516594
Creatinine113.128.4840.075
D-Glucose (Dextrose)180.16900.85.0
Fructose180.167.2060.03999778
Galactose180.1610.810.060002223
Glycerol92.0911.050.11999132
Hypoxanthine136.111.36110.01
Phenol Red376.45.270.014001063
Urea60.06300.34.9999995
Uridine244.20.7330.003001638
Non-Proteinogenic Amino Acids
Acetylcarnitine239.71.1990.005002086
Betaine117.158.2010.07000427
L-Amino-N-Butyric Acide103.122.0620.019996122
L-Carnitine HCl198.07.920.04
L-Citrulline175.27.0080.04
L-Hydroxyproline131.132.6230.020003049
L-Ornithine168.6211.8030.06999763
N-Acetylglycine117.110.5390.089999996
Taurine125.1411.2630.09000319
Water-Soluble Acids
3-Hydroxybutyric Acid Sodium126.096.3050.050003964
Alpha Ketoglutaric Acid146.10.7310.005003422
Citric Acid Anhydrous192.1324.9770.13000052
Formic Acid46.032.3010.04998914
Malic Acid134.090.670.0049966443
Malonic Acid104.061.0410.010003844
Sodium Acetate Anhydrous82.033.2810.039997563
Sodium L-Lactate112.06179.2961.6
Sodium Pyruvate110.045.5020.049999997
Succinic Acid118.092.3620.020001695
Uric Acid168.1158.83850.35
Reference:
Related Products: