Search
HeLa cells were loaded with 200 nM TMRM in Live Cell Imaging Solution for 30 minutes at 37°C. Cells were washed and then loaded with CellMask™ Green Plasma Membrane Stain for 15 minutes at room temperature. Images were acquired on a confocal microscope.