Immunocytochemistry analysis of ABfinity™ Recombinant ZO-1 Rabbit Oligoclonal Antibody
Immunocytochemistry analysis of Caco-2 stained with ABfinity™ Recombinant ZO-1 Rabbit Oligoclonal Antibody (2µg/mL) using a: Alexa Fluor® 488 goat anti-rabbit as a secondary antibody (green). b: DAPI stained Caco-2 cell nuclei (blue). c: Composite image of cells showing antibody staining membrane and tight junction. d: no primary control.
Immunocytochemistry analysis of Caco-2 stained with ABfinity™ Recombinant ZO-1 Rabbit  Oligoclonal Antibody (2µg/mL) using a: Alexa Fluor® 488 goat anti-rabbit as a secondary antibody (green). b: DAPI stained Caco-2 cell nuclei (blue). c: Composite image of cells showing antibody staining membrane and tight junction. d: no primary control.
Related Products
Related Images