• Each gel contains 48 or 96 sample lanes and 8 marker lanes that are ideal for screening multiple PCR products.
  • The unique 96-well staggered-well format provides a 1.6-cm run length that resolves between 50 bp and 10 kb.
  • The E-Gel 96 loading format is compatible with multi-channel pipettors, and the most commonly used 8-, 12-, and 96-pin liquid handling robots.
  • With 12-minute run times, as opposed to ~1 hour-30 minute preparation plus 20 minute running + slow loading, up to 20,000 samples can be resolved in a single day.
E-Gel frees up space, routine work time, disposal concerns, and increases safety.