SeCore DRB1 Locus Exon 2 & 3 Sequencing Kit
Now available: the new SeCore DRB1 Locus Exon 2 & 3 Sequencing Kit offers complete bi-directional sequencing of exon 2 & exon 3, including the first hypervariable region
   See the results. Watch the video.
The complete bidirectional sequencing of DRB1 exon 2 and exon 3 is designed to provide clean and clear sequencing data, including the first hypervariable region. The HLA research and development team at Life Technologies formulated new proprietary amplification and sequencing primers outside exon 2 and exon 3, and the result is easy to read forward and reverse sequencing data at more positions.
Labs adopting this new SeCore DRB1 exon 2 and exon 3 kit can have greater confidence in results and a reduction in risk of misinterpretation because the DRB1 sequencing data offers complete bi-directional exon coverage. Improved sequencing data can reduce the occurrence of secondary verification methods, which can help reduce labor costs and provide a shorter time to a final typing result. All SeCore kits include the enzymes and BigDye chemistry reagents needed for the workflow, relieving your staff from spending extra time on quality-control steps.
Learn more about the resolution capability
Click the button below to complete a short form and a Life Technologies HLA technical sales specialist will contact you to discuss details of how this new kit differs from others currently available, and to share examples of the bi-directional sequencing data that can be expected from the new SeCore DRB1 kit.
Contact sales specialist