The Identifiler Direct kit dramatically simplifies the workflow for the processing of single source samples. This chemistry was developed specifically to eliminate the need for extraction and purification of blood or buccal samples on FTA paper, enabling users to just "punch and go."