• A single amplification provides information on the eight largest and most difficult to amplify loci in the AmpFℓSTR Identifiler and SGM Plus PCR Amplification Kits including D13S317, D7S820, D2S1338, D21S11, D16S539, D18S51, CSF1PO and FGA
  • Loci are compatible with CODIS and international databasing standards
  • Includes amplification of amelogenin allowing sex determination
  • Utilizes proven 5-dye technology allowing compatibility of workflows with Identifiler, SEfiler™ and Yfiler kits
  • Optimized for use with the most difficult types of samples including those that are degraded and contain inhibitors
  • Ability to overcome the inhibitory effects of substances such as heme and humic acid which are commonly encountered in forensic samples
  • Can be used as an adjunct to other commercially available amplification kits to recover the larger loci, which cannot be amplified or that drop out
  • Recovery of full profiles with 125 pg input DNA
  • Highly discriminating results with Probability of Identity values of less than 1 X 10-10 for US populations
  • Developmental validation studies performed according to the Scientific Working Group on DNA Analysis Methods (SWGDAM) guidelines
  • Please see the AmpFℓSTR Kit Product Portfolio, showing loci amplified by kit, dye label and probabilities of identity. (93 KB PDF)