As the world's first commercially available miniSTR kit, the AmpFℓSTR MiniFiler™ PCR Amplification Kit increases your ability to obtain DNA results from compromised samples that previously would have yielded limited or no genetic data. This means cold cases can come off the shelf for re-analysis and new, challenging samples have a better chance of delivering interpretable results.