GIBCO Protein Expression Medium (PEM)
  • Supports the large-scale, high-density culture of transfected PER.C6 cells in shake flasks, roller bottles, and bioreactors
  • Contains no components directly derived from human or animal sources and one low-level, recombinant protein, minimizing the opportunity to introduce adventitious agents
  • Formulated without L-glutamine for maximum stability
  • Supports parental PER.C6, HEK 293, and HeLa cell growth in suspension culture, as well as transfected PER.C6 cells for protein expression applications