高保真PCR酶
对于序列准确度极为关键的应用领域,需要利用高保真PCR或降低错误率。 To obtain high fidelity, proofreading enzymes with 3’ to 5’ exonuclease activity, such as Platinum Taq High Fidelity are included in our PCR mixes. We offer a range of products to give you the fidelity you need for your applications. To make it easy to choose your product, we suggest to first choose based on your need for fidelity, then on the target length. If your application also calls for additional specificity, then we recommend one of the AccuPrime™ products.
Which High Fidelity PCR Enzyme Is Right for You?
  Basic High Fidelity Choice for Long Template Choice for Long Template with Enhanced Fidelity & Specificity Preferred high Fidelity Choice Ultimate Choice for High Fidelity with Enhanced Specificity Special High Fidelity Choice for Small Templates
畅销产品是的 
保真度 6x 9x 26x 26x 50x
Enhanced Specificity是的是的
Amplicon Size >20 kb >20 kb >20 kb >12 kb >4 kb
High Yield 是的是的
Master Mix/Super Mix Format是的 是的
立即订购立即订购立即订购立即订购 立即订购