Affinity determination of Rabbit Anti-Goat IgG (H+L) Superclonal™ Secondary Antibody, unconjugated
Histogram showing the affinity (Kd) of the Rabbit Anti-Goat IgG (H+L) Superclonal Secondary Antibody, unconjugated (A27011) to goat IgG. Based on kinetic characterization of superclonal antibody using the Octet system (ForteBio), the affinity of three lots of the superclonal antibody was consistently in the low picomolar (pM) range.
Histogram showing the affinity (Kd) of the Rabbit Anti-Goat IgG (H+L) Superclonal Secondary Antibody, unconjugated (A27011) to goat IgG. Based on kinetic characterization of superclonal antibody using the Octet system (ForteBio), the affinity of three lots of the superclonal antibody was consistently in the low picomolar (pM) range.
Related Products
Related Images