PG concentration in human serum
The figure shows the PG concentration in serum of normal subjects aged from 3 to 66 years. The PG levels measured in adult sera (age: 16-66 years) ranged between 1 and 4.4 µg/ml and the mean concentration was 2.8 µg/ml. PG levels measured in children sera (age: 3-16 years) ranged between 7 and 15 µg/ml. The mean PG concentration was 10.4 µg/ml.
The figure shows the PG concentration in serum of normal subjects aged from 3 to 66 years. The PG levels measured in adult sera (age: 16-66 years) ranged between 1 and 4.4 µg/ml and the mean concentration was 2.8 µg/ml. PG levels measured in children sera (age: 3-16 years) ranged between 7 and 15 µg/ml. The mean PG concentration was 10.4 µg/ml.
Related Products
Related Images