Desktop SEM with a durable electron source and exceptional image quality
Available in three configurations to take you from basic tasks to advanced analyses, Thermo Scientific Phenom G6 Desktop SEMs are fitted with cerium hexaboride (CeB6) thermionic electron sources. These durable electron sources provide exceptional image quality with improved resolution and contrast, lasting up to 30 times longer than traditional tungsten filaments. Unlike tungsten filaments, CeB6 sources degrade gradually rather than experiencing sudden failures, so you can plan replacements during regular maintenance.
Phenom Pure G6 Desktop SEM: Suited to basic applications
The Phenom Pure G6 Desktop SEM is a basic tabletop SEM suited for educational settings and standard research and development tasks. It’s a straightforward imaging solution with a focus on ease of use and affordability.
This entry-level SEM performs better than an optical microscope. In fact, thanks to its reliable CeB6 source, the Phenom Pure G6 Desktop SEM delivers higher resolution than other systems in its class. Just 10 minutes of training is all that’s required to begin using this microscope, with many key functions automated to save time and effort.  
  • Exceptional image quality thanks to the high-brightness, long-lasting CeB6 source
  • User-friendly design makes operation intuitive for both beginners and experienced users
  • Outstanding cost-to-benefit ratio makes high-quality imaging more accessible
  • Upgradability allows you to level up on site to the Phenom Pro or ProX Desktop SEM models for enhanced capabilities
Uncoated wood imaged with a Phenom Pure G6 Desktop SEM
Lily pollen imaged with a Phenom Pure G6 Desktop SEM
Paracetamol imaged with a Phenom Pure G6 Desktop SEM
Red rust imaged with a Phenom Pure G6 Desktop SEM
Phenom Pro G6 Desktop SEM: Advanced imaging performance
The Phenom Pro G6 Desktop SEM elevates performance by providing a comprehensive imaging system suitable for a variety of advanced applications.
The Phenom Pro G6 Desktop SEM delivers high-quality results from a compact and practical desktop format. Its efficient sample loading mechanism enables faster and more productive routine analysis with minimal time spent tuning between experiments.  
  • High-brightness CeB6 source delivers outstanding image resolution across various accelerating voltages
  • Quick operation enables electron imaging in just 30 seconds from sample loading
  • Intuitive design ensures seamless operation for both novice and experienced users
  • Upgradability allows you to level up on site to the Phenom ProX Desktop SEM for enhanced capabilities
Enhanced features:
  • Multiple acceleration voltages ranging from 5.0 to 20.5 kV
  • Color optical camera for quick navigation
  • Charge reduction mode minimizes the need for sample coating
  • Optional secondary electron detector
  • Compact design requires no additional infrastructure
  • Low maintenance and high stability with vibration isolation
  • Various sample holders for optimized imaging
White coral imaged with a Phenom Pro G6 Desktop SEM
Diatom imaged with a Phenom Pro G6 Desktop SEM
Mosquito proboscis imaged with a Phenom Pro G6 Desktop SEM
Coated nanoparticle imaged with a Phenom Pro G6 Desktop SEM
Phenom ProX G6 Desktop SEM: Comprehensive imaging and EDS analysis
For advanced analytical capabilities, the Phenom ProX G6 Desktop SEM integrates seamlessly with an energy dispersive X-ray spectroscopy (EDS) detector to provide detailed X-ray elemental analysis in addition to high-performance imaging.
With its integrated EDS detector, the Phenom ProX G6 Desktop SEM is a total research solution right on your desk—including seamless elemental analysis. Like all models in the Phenom G6 Desktop SEM family, it’s compact, fast, and easy to use, delivering high-resolution images with outstanding brightness.  
Enhanced features:
  • Includes all the advanced imaging capabilities of the Phenom Pro G6 Desktop SEM
  • Integrated EDS detector provides detailed X-ray elemental analysis for comprehensive material characterization
  • User-friendly interface offers smooth operation, even for those new to SEM
  • Enhanced analytical capabilities support applications that require detailed elemental analysis and imaging
Sandpaper imaged with a Phenom ProX G6 Desktop SEM
Stainless steel imaged with a Phenom ProX G6 Desktop SEM
Ink on paper imaged with a Phenom ProX G6 Desktop SEM
Plaster imaged with a Phenom ProX G6 Desktop SEM
Phenom G6 Desktop SEM selection guide
The Phenom Pure, Pro, and ProX G6 Desktop SEMs provide a scalable solution for your imaging needs, from the cost-effective Phenom Pure G6 Desktop SEM to the high-performance Phenom Pro G6 Desktop SEM and the analytically advanced Phenom ProX G6 Desktop SEM. Choose the model that best fits your requirements and enjoy the benefits of exceptional imaging, ease of use, and upgrade flexibility.
Use this comparison to understand the key differences between the Phenom Pure, Pro, and ProX G6 Desktop SEMs. It highlights the unique features and capabilities of each model, allowing you to choose the system that best fits your imaging and analytical needs.
  Phenom Pure G6 Desktop SEM
Phenom Pro G6 Desktop SEM Phenom ProX G6 Desktop SEM
Acceleration voltages Multiple voltages (5 to 10 kV) Multiple voltages (5 to 20.5 kV) Multiple voltages (5 to 20.5 kV)
Charge reduction mode No Yes Yes
Secondary electron detector No Optional Optional
Upgrade path Upgradable to Pro and ProX SEMs Upgradable to ProX SEM N/A
Technical specifications for Phenom Pure, Pro, and ProX G6 Desktop SEMs
  Phenom Pure G6 Desktop SEM Phenom Pro G6 Desktop SEM Phenom ProX G6 Desktop SEM
Sample size
  • Up to 25mm diameter (optionally up to 32mm)
  • Up to 35mm height (optionally up to 100mm)
  • Up to 25mm diameter (optionally up to 32mm)
  • Up to 35mm height (optionally up to 100mm)
  • Up to 25mm diameter (optionally up to 32mm)
  • Up to 35 mm height (optionally up to 100 mm)
Source type Cerium hexaboride (CeB6) Cerium hexaboride (CeB6) Cerium hexaboride (CeB6)
Max. resolution  < 15nm  ≤ 6nm ≤ 6nm
Detectors and signals
  • Backscattered electron detector (standard)
  • EDS detector (optional)
  • Everhart-Thornley (ETD) SE detector (optional)
  • Backscattered electron detector (standard)
  • EDS detector (optional)
  • Everhart-Thornley (ETD) SE detector (optional)
  • Backscattered electron detector (standard)
  • EDS detector (standard)
  • Everhart-Thornley (ETD) SE detector (optional)
Software options and accessories
  • ChemiSEM Technology
  • PPI/PPA
  • ProSuite
  • Maps 3 Software
  • Avizo Software
  • Avizo Trueput Software
  • ChemiSEM Technology
  • PPI/PPA
  • ProSuite
  • Maps 3 Software
  • Avizo Software
  • Avizo Trueput Software
  • ChemiSEM Technology
  • PPI/PPA
  • ProSuite
  • Maps 3 Software
  • Avizo Software
  • Avizo Trueput Software
Footprint
286(w) x 566(d) x 495(h) mm, 50 kg
286(w) x 566(d) x 495(h) mm, 50 kg
286(w) x 566(d) x 495(h) mm, 50 kg
Acceleration voltage range 
5 kV to 15 kV
5 kV to 15 kV
Advanced mode: Adjustable between 4.8 kV and 20.5 kV 
5 kV to 15 kV
Advanced mode: Adjustable between 4.8 kV and 20.5 kV 
Low vacuum mode (stating pressure ranges) 1 Pa – 60 Pa  1 Pa – 60 Pa 1 Pa – 60 Pa
For Research Use Only. Not for use in diagnostic procedures.