Search
With electron microscopy, we can see breakthroughs everywhere, whether it’s improving the food we grow, understanding what keeps us healthy, or developing new therapeutics that help us when things go wrong. The Thermo Scientific Talos 12 Transmission Electron Microscope (TEM) is the next step in our 120 kV instrumentation, designed to fit your needs.
The user-friendly Talos 12 TEM offers exceptional performance for a diverse range of scientific imaging and experimentation needs. It features a reduced footprint with a robust enclosure, along with systems that improve accessibility, bringing trusted performance and automation to more researchers and their labs.
It can be configured as a conventional system for routine TEM imaging of cells and tissues, as an advanced system for sample characterization with different scanning transmission electron microscopy (STEM) and energy-dispersive X-ray spectroscopy (EDS) detectors, or even as an entry-level cryo-electron microscopy (cryo-EM) platform. Its modular design allows it to meet the diverse needs of both research labs and core facilities.
For Research Use Only. Not for use in diagnostic procedures.