A streamlined, reliable, and consistent combustion ion chromatography platform
The Thermo Scientific Cindion Combustion Ion Chromatography (C-IC) System is thoughtfully designed to simplify your combustion IC analysis, while providing you with the flexibility to expand your application capabilities as needed. Explore the features that make the Cindion C-IC System stand out.
Designed for PFAS analysis
Through its innovative design, the Cindion C-IC System minimizes use of materials that can contribute to detectable PFAS levels and achieves exceptionally low background PFAS levels. The system design enables highly sensitive PFAS detection, making it suitable for methods like US EPA Method 1621.
Enhanced screening of PFAS compounds in wastewater: Implementing US EPA Method 1621 with improved C-IC
Fully controlled by a single software
The Cindion C-IC System can be controlled completely by the Thermo Scientific Chromeleon Chromatography Data System (CDS). Simply set up a single sequence to control the combustion and data analysis process, from sample selection to report generation.
Small system footprint
The Cindion C-IC System footprint has a minimal impact on the space in your lab. The novel, compact combustion tube design ensures complete combustion in a shorter distance. Additionally, the autosampler and absorption tube are integrated into a single module.
Industry-leading IC system
The Cindion C-IC System takes advantage of the features and benefits of the Thermo Scientific Dionex Inuvion IC System. With innovations such as electrolytic suppression and Reagent-Free IC (RFIC) eluent generation, the Cindion C-IC System provides consistent IC analyses in a more user-friendly package.
The LC-MS are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks. They are designated for Research Use Only Applications.
연구용으로만 사용 가능합니다. 진단용으로는 사용할 수 없습니다.