Search
Find out more about our pre-installed software, firmware, cloud solutions, and drivers, as well as access to our downloads.
Our dEX platform comes pre-installed on every logger in the DT80 range.
dEX 2.0 is a simplified yet powerful software that allows you to configure and program your DT80 series data loggers. This configuration-based software allows you to configure the hardware and visualisation data and also debug your application on all DT80 series of loggers (including DT82 and DT85 series).
Core enhancements
Hardware support
Hardware support is available for the DT80 series 3 and 4.
![]() | ![]() |
Homepage
| Setting Wizard
|
![]() | ![]() |
Data Explorer
| Dashboard
|
DataTaker Live is built to help companies collect, monitor, analyze, store, and share information that improves connectivity and data acquisition of process instruments and industrial systems. Our solution increases operational productivity and efficiency.
Monitor—Gain real-time remote access to measurements from anywhere at any time and allowing configuration changes for dataTaker loggers from your computing devices.
Store & Share—Synchronize and save data to your personal cloud account. dataTaker Live dashboard lets you collaborate and share data.
Collect—Automatic data uploading from sensors, loggers, or Industrial gateways to your personal cloud service account.
Analyze—Data analysis with standard and custom scripts.
The DataTaker Live dashboard displays real-time sensor measurements, calculations, and diagnostic information using a custom dashboard.
DataTaker Live raises alarms to alert users through the interface using color coding when an abnormal condition or problem is detected.
Please read the following notes prior to downloading firmware.
| Firmware | Modem Firmware | Host Software |
The dataTaker LabVIEW Driver is a comprehensive library of VIs that allows the dataTaker range of data loggers to be configured and queried from LabVIEW.
Features:
Use the driver VIs to take care of all the communications and data formatting of the dataTaker data loggers, allowing you to concentrate on your application rather than how to interface to the data logger.
With flexible, universal analog, digital, and serial inputs, the dataTaker hardware can be used for real-time data acquisition, in addition to being perfect for applications requiring stand-alone operation in remote or distributed locations with limited power supply.
The dataTaker DT8x driver for LabVIEW has been written to interface with the DT80 data loggers. Download the driver, and extract the zip file to the relevant user.lib directory in the LabVIEW directory.
The DataTaker Instrument driver for LabVIEW has been written to interface to the DT800 data loggers and the DT5xx range. Download the driver and extract the zip file to the relevant user.lib directory in the LabVIEW directory.
dEX 2.0 is a simplified yet powerful software that allows you to configure and program your DT80 series data loggers. This configuration-based software allows you to configure the hardware and visualisation data and also debug your application on all DT80 series of loggers (including DT82 and DT85 series).
Core enhancements
Hardware support
Hardware support is available for the DT80 series 3 and 4.
![]() | ![]() |
Homepage
| Setting Wizard
|
![]() | ![]() |
Data Explorer
| Dashboard
|
DataTaker Live is built to help companies collect, monitor, analyze, store, and share information that improves connectivity and data acquisition of process instruments and industrial systems. Our solution increases operational productivity and efficiency.
Monitor—Gain real-time remote access to measurements from anywhere at any time and allowing configuration changes for dataTaker loggers from your computing devices.
Store & Share—Synchronize and save data to your personal cloud account. dataTaker Live dashboard lets you collaborate and share data.
Collect—Automatic data uploading from sensors, loggers, or Industrial gateways to your personal cloud service account.
Analyze—Data analysis with standard and custom scripts.
The DataTaker Live dashboard displays real-time sensor measurements, calculations, and diagnostic information using a custom dashboard.
DataTaker Live raises alarms to alert users through the interface using color coding when an abnormal condition or problem is detected.
Please read the following notes prior to downloading firmware.
| Firmware | Modem Firmware | Host Software |
The dataTaker LabVIEW Driver is a comprehensive library of VIs that allows the dataTaker range of data loggers to be configured and queried from LabVIEW.
Features:
Use the driver VIs to take care of all the communications and data formatting of the dataTaker data loggers, allowing you to concentrate on your application rather than how to interface to the data logger.
With flexible, universal analog, digital, and serial inputs, the dataTaker hardware can be used for real-time data acquisition, in addition to being perfect for applications requiring stand-alone operation in remote or distributed locations with limited power supply.
The dataTaker DT8x driver for LabVIEW has been written to interface with the DT80 data loggers. Download the driver, and extract the zip file to the relevant user.lib directory in the LabVIEW directory.
The DataTaker Instrument driver for LabVIEW has been written to interface to the DT800 data loggers and the DT5xx range. Download the driver and extract the zip file to the relevant user.lib directory in the LabVIEW directory.