Search
| Affordable & pure, flow compatible cells | Highest purity for molecular assays | Untouched—most physiological relevant cells | |
|---|---|---|---|
| Dynabeads FlowComp Mouse CD4 | Dynabeads Mouse CD4 | Dynabeads Untouched Mouse CD4 Cells | |
| Isolation method | Positive isolation | Depletion or positive isolation | Negative isolation |
| Starting samples | Any sample (whole blood, spleen, lymph node) | Any sample (whole blood, spleen, lymph node) | Spleen, lymph node cells* |
| # Cells processed | 2 x 109 cells | 1-2 x 109 cells | 1 x 109 cells |
| Purity | Higher 90-98% | Higher 90-98% | High 80-90% |
| Yield | Standard 50-60% | Highest 95-100% | Standard 50-70% |
| Viability | >90% | >90% | >90% |
| Isolation time | ~60min | ~30 min | ~50 min |
| Isolation temperature | cold | cold | cold |
| Applications | Cellular assays (cell culture, cell activation/proliferation, cytokine studies) Molecular: isolation of proteins & nucleic acids | Depletion of unwanted CD4+ T cells or Positive isolation for molecular isolation of proteins & nucleic acids | Cellular applications for physiological studies (functional studies, functional studies, adoptive transfer, flow analysis or further sorting) |
| Bead-free target cells | Yes (but antibody present) | No | Yes |
| Compatible with flow cytometry /flow sorting | ![]() | ![]() | |
| Shelf life from production date | 2 years | 3 years | 3 years |
| Required materials | Magnets | Magnets | Magnets |
| Order Now | Order Now | Order Now |
THIS IS FOR MOUSE RESEARCH - NOT FOR HUMAN
For Research Use Only. Not for use in diagnostic procedures.