Search
Primary XPS region: Ar2p
Overlapping regions: N/A
Binding energies of common chemical states:
| Chemical state | Binding energy Ar2p |
|---|---|
| Implanted Ar | ~243 eV |
Experimental information
Interpretation of XPS spectra
About this element
Symbol: Ar
Date of discovery: 1894
Name origin: Greek argos
Appearance: colorless gas
Discoverer: Ramsay, Rayleigh
Obtained from: air
Melting point: 83.8 K
Boiling point: 87.3 K
Density[kg/m3]: 1.784
Molar volume: 22.56 × 10-6 m3/mol
Protons/Electrons: 18
Neutrons: 22
Shell structure: 2,8,8
Electron configuration: [Ne]3s23p6
Oxidation state: 0
Crystal structure: cubic face centered
Argon was discovered in 1894 by Lord Rayleigh and Sir William Ramsay, although Henry Cavendish suspected its existence over one hundred years earlier. This noble gas is chemically inert and is colorless and odorless in its liquid and gaseous forms. Argon makes up slightly less than 1% of the Earth’s atmosphere. There are no known stable chemical compounds containing argon. Due to its inert nature, argon is used in lighting, as a blanket in the manufacture of titanium, and in other applications needing a nonreactive medium.
Products
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.