Search
Primary XPS region: N/A
Overlapping regions: N/A
Binding energies of common chemical states: N/A
Experimental information
Interpretation of XPS spectra
N/A
General comments
About this element
Symbol: He
Date of discovery: 1895
Name origin: Greek helios
Appearance: colorless
Discoverer: Sir William Ramsay
Obtained from: natural gas deposit air
Melting point: 1.15 K
Boiling point: 4.55 K
Density[kg/m3]: 0.1785
Molar volume: 21.0 × 10-6 m3/mol
Protons/Electrons: 2
Neutrons: 2
Shell structure: 2
Electron configuration: 1s2
Oxidation state: 0
Crystal structure: hexagonal
While helium was first detected in 1868 as a bright yellow line in the solar spectrum, it took until 1895 for W. Ramsay to isolate it from cleveite. In 1905, H. Cady and D. McFarland isolated helium from natural gas, the current source for most helium today. Helium is a colorless, odorless gas under standard conditions. However, it does exhibit an unusual liquid state near absolute zero known as superfluidity. Many are aware of helium’s lifting power for airships and balloons. Helium is also used as a coolant in MRI systems, superconducting magnets and cryogenics because of its low melting and boiling points. Helium’s inert properties also allow it to be used as a coolant for nuclear reactors.
Products
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.