Bismuth • Other Metal
Primary XPS region: Bi4f
Overlapping regions: Si2s loss features
Binding energies of common chemical states:
Chemical stateBinding energy Bi4f7/2
Bi metal157 eV
Bi2O3159 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Bi4f region has well-separated spin-orbit components (Δ=5.3eV).
    • Peaks in the Bi4f region have an asymmetric peak shape for bismuth metal.
    • Bismuth compounds, such as its oxide, have symmetric Bi4f peaks.
element-bismuth-xpsspectra
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
element_bismuth
About this element
Element Crystal rhom
Symbol: Bi
Date of discovery: unknown
Name origin: German wissmuth
Appearance: white
Discoverer: unknown
Obtained from: bismuthine
Melting point: 271.3 K
Boiling point: 1560 K
Density[kg/m3]: 9.8
Molar volume: 21.31 × 10-6 m3/mol
Protons/Electrons: 83
Neutrons: 26
Shell structure: 2,8,18,32,18,5
Electron configuration: [Xe]4f145d106s26p3
Oxidation state: 3,5
Crystal structure: Rhombohedral
This brittle, pinkish metal gets its name from the German word wissmuth, meaning white mass. In early times, bismuth was often confused with tin or lead due to its resemblance to those elements. When bismuth is combusted with oxygen, it burns blue and its oxide forms yellow fumes. Bismuth is used in cosmetics. It is found in pharmaceuticals and is used to treat gastrointestinal diseases. Strong permanent magnets are produced using one of its alloys, and bismuth has been used as a dense material in fishing sinkers and as a replacement for lead in shot and bullets.
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards