Lead • Other Metal
Primary XPS region: Pb4f
Overlapping regions: P2p, Zn3s
Binding energies of common chemical states:
Chemical stateBinding energy Pb4f7/2
Pb metal136.9 eV
PbO2137.8 eV
Pb3O4138.4 eV
Pb native oxide138.4 eV
2PbCO3.Pb(OH)2138.4 eV
Pb palmitate138.4 eV
Pb azelate138.4 eV
Native oxide referenced to Pb metal. All others charge referenced to adventitious C1s peak at 284.8eV.
Experimental information
N/A
Interpretation of XPS spectra
  • Pb4f region has well-separated spin-orbit components (Δmetal=4.87eV).
    • Not always a valid charge reference value (e.g., C1s peak for adventitious carbon on native oxide of aluminum appears at ~286eV).
  • Analysis of the O1s region for air-exposed Pb metal indicates the native oxide has a high concentration of hydroxide.
element-lead-xpsspectra
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
element_lead
General comments
  • PbO2 may have a small amount of PbCO3 formed on the surface through reaction with CO2 in the atmosphere.
element-lead-xpsspectra2
About this element
Element Crystal cub
Symbol: Pb
Date of discovery: known to the ancients
Name origin: Latin plumbum
Appearance: bluish-white
Discoverer: unknown
Obtained from: galena
Melting point: 600.1 K
Boiling point: 2022 K
Density[kg/m3]: 11.340
Molar volume: 18.26 × 10-6 m3/mol
Protons/Electrons: 82
Neutrons: 125
Shell structure: 2,8,18,32,18,4
Electron configuration: [Xe]4f145d106s26p2
Oxidation state: 4,2
Crystal structure: cubic
This blue-white metal is soft and malleable. A poor conductor of electricity, lead is resistant to corrosion and very dense. Lead derives its symbol Pb from the Latin word plumbum, meaning “liquid silver.” Prolonged exposure to lead causes neurological damage as well as renal disease, cardiovascular effects, and reproductive toxicity. Although the harmful effects of lead have been recognized as early as 2000 BC, it has been used throughout history for plumbing and ammunition. Recently, lead has been used in storage batteries, paints, petrol, and sound absorbance materials, although these uses are being phased out due to health and environmental effects.
Products
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.