Platinum • Transition Metal
Primary XPS region: Pt4f
Overlapping regions: Al2p
Binding energies of common chemical states:
Chemical stateBinding energy Pt4f7/2
Pt metal71.0 eV
PtO72.4 eV
PtO274.9 eV
Interpretation of XPS spectra
  • Pt4f region has well separated spin-orbit components (Δmetal=3.35eV).
    • Peaks in the Pt4f region have an asymmetric peak shape for platinum metal.
    • Platinum compounds, such as the oxide, have symmetric Pt4f peaks.
About this element
Element Crystal cub
Symbol: Pt
Date of discovery: 1735
Name origin: Spanish platina
Appearance: silver
Discoverer: Julius Scaliger
Obtained from: platinum ores
Melting point: 2041 K
Boiling point: 4098 K
Density[kg/m3]: 21090
Molar volume: 9.09 × 10-6 m3/mol
Protons/Electrons: 78
Neutrons: 117
Shell structure: 2,8,18,32,17,1
Electron configuration: [Xe]4f145d96s1
Oxidation state:  2,4
Crystal structure: cubic
Discovered by J. Scaliger in 1735, platinum is a beautiful, gray-white metal that is malleable in its pure form. It is extremely resistant to corrosion and tarnishing, and will not oxidize in air, even at high temperatures. Platinum is very rare, and is naturally found as the uncombined metal. Because of its extreme durability and resistance to tarnishing, platinum is widely used to make jewelry. It is also used to make surgical tools, liquid crystal displays, and electrical resistance wires. Platinum is named for the Spanish word “platina,” meaning “little silver.”
element_platinum
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.