Growth of frozen Chlamydomonas reinhardtii
Three Chlamydomonas reinhardtii cultures were grown at 50 µE m-2s-1, 26°C, and 110 rpm, and optical densities were measured at OD750 at 16, 24, 40, 48, 64, and 80 hrs.  The culture represented by the blue line started with a fresh culture, and the cultures represented by the red and green lines were frozen at different cell concentrations using the GeneArt® Cryopreservation Kit for Algae and resuscitated in TAP growth media, following manufacturer recommendations.   Similar results were seen with Chlorella vulgaris, either wild type or transformants.
Three Chlamydomonas reinhardtii cultures were grown at 50 µE m<sup>-2</sup>s<sup>-1</sup>, 26°C, and 110 rpm, and optical densities were measured at OD750 at 16, 24, 40, 48, 64, and 80 hrs.  The culture represented by the blue line started with a fresh culture, and the cultures represented by the red and green lines were frozen at different cell concentrations using the GeneArt® Cryopreservation Kit for Algae and resuscitated in TAP growth media, following manufacturer recommendations.   Similar results were seen with Chlorella vulgaris, either wild type or transformants.
Related Products
Related Images