ICSR supplementation is equivalent to human serum for CAR engineered T cell expansion
Primary human T cells were obtained from the University of Pennsylvania Human Immunology Core. T cells were activated with Dynabeads CD3/CD28 at a 3:1 bead per cell ratio in the respective media (commercially available hematopoietic medium with 5% human AB serum or 5% Immune Cell Serum Replacement [ICSR]) and transduced with CART19 lentiviral vector. Cells were expanded in T-225 flasks over the duration of the culture. T cells were counted with a Coulter Multisizer 3 and expanded until they reached near resting size. T cell expansion is represented as cumulative T cell numbers plotted over time.
Acknowledgement
We thank James L. Riley and Andrew Medvec from the University of Pennsylvania, Department of Microbiology, for their support in generating this data.
Primary human T cells were obtained from the University of Pennsylvania Human Immunology Core. T cells were activated with Dynabeads CD3/CD28 at a 3:1 bead per cell ratio in the respective media (commercially available hematopoietic medium with 5% human AB serum or 5% Immune Cell Serum Replacement [ICSR]) and transduced with CART19 lentiviral vector. Cells were expanded in T-225 flasks over the duration of the culture. T cells were counted with a Coulter Multisizer 3 and expanded until they reached near resting size. T cell expansion is represented as cumulative T cell numbers plotted over time.<br><br><b>Acknowledgement</b><br>We thank James L. Riley and Andrew Medvec from the University of Pennsylvania, Department of Microbiology, for their support in generating this data.
Related Products
Related Images