Search
BODIPY® 630⁄650-X SE (Cat. no. D10000) applied to a Pseudomonas aeruginosa biofilm. BODIPY® 630⁄650-X SE binds to a Pseudomonas aeruginosa PAO1 pMF230 biofilm matrix. The image was obtained using a Leica TCS-SP2 AOBS confocal microscope and a 63x⁄0.9 NA water immersion objective. Image contributed by Betsey Pitts, Microscopy Center for Biofilm Engineering, Bozeman, Montana.