Table of Contents
Component
e204 nBos d 6, BSA Cow
e204 nBos d 6, BSA Cow Scientific Information
Type:
Component
Name; WHO/IUIS:
nBos d 6, BSA Cow
Biological function:
Bovine serum albumin
Allergen code:
e204
Source Material:
nBos d 6 is purified from a cow extract
Other Names :
Bovine serum albumin; BSA
Allergen
Cow's milk (CM) is popularly consumed throughout the world. Allergy to CM is prevalent among infants and young children during the first year of life, affecting 0.5-3% at the age of 1 year. Bos d 6 (bovine serum albumin) has been reported as a minor allergenic whey protein in CM. According to a number of studies, sensitization towards Bos d 6 ranges between 43-92.9% of patients with CM allergy (CMA). Bos d 6 is the most heat-labile protein among the other whey proteins and therefore is likely to be destroyed after boiling CM. Thus, specific immunoglobulin E to Bos d 6 might help evaluate tolerance to boiled milk. Clinical manifestations of Bos d 6-mediated allergy majorly include anaphylaxis. Moreover, sensitization to Bos d 6 is regarded as a predictive marker of CMA in children with beef allergies. Cross-reactivity has been observed between serum albumin of cow, deer, dog, sheep, pork, and cat
Epidemiology
Worldwide distribution
Cow’s milk (CM) is a popular food consumed worldwide (Muthukumar et al. 2020). However, CM allergy (CMA) is prevalent among infants and young children, with a high prevalence observed during the first year of life (Kaczmarski et al. 2013, Vandenplas 2017), affecting 0.5-3% at the age of 1 year (Flom et al. 2019, Skripak et al. 2007).
Bos d 6 is bovine serum albumin (BSA) and a minor allergenic whey protein in CM. According to a study, 43% of patients with CMA (n=92) are found to be sensitized to Bos d 6 (Hochwallner et al. 2014, Wal 2002). According to another clinical study, 61.3% of children allergic to CM (n=80) reported sensitization towards Bos d 6 (based on immunoblotting) (Restani et al. 1999).
In an Italian in-vitro study, 45% (9/20) of pediatric patients allergic to CM exhibited sensitization to Bos d 6 (Natale et al. 2004). As per another study conducted in Italy, sensitization to Bos d 6 was observed in 92.9% (26/28) of the children allergic to CM (Martelli et al. 2002).
Furthermore, specific immunoglobulin E (sIgE) against Bos d 6 was reported among 45.5% of patients with CMA (n=86) in an Iran-based study (Shoormasti et al. 2011). And in a study conducted in Denmark among 39 milk-allergic infants [21 infants with IgE-CMA (CMA) and 18 infants with non-IgE CMA (CMI)], the frequency of sIgEs to Bos d 6 was highest before (12/17 for CMA; 9/13 for CMI) and after (15/20 for CMA; 15/17 for CMI) the milk challenge at 12 months in comparison to other cow milk proteins (Host et al. 1992).
Moreover, a study conducted in Europe reported the presence of sIgE towards Bos d 6 in 3.8% of the patients (children and adults) with CMA (n=73) (Hochwallner et al. 2010). A study used crossed radioimmunoelectrophoresis technique to determine the serum IgE level to CM antigenic components in the sera of 21 CMA patients and detected IgE antibodies against bovine albumin in the sera of all patients (Gjesing et al. 1986).
Environmental Characteristics
Source and tissue
Cow milk comprises two main types of proteins, whey (constituting about 20% of the total milk protein) and casein (constituting about 80% of the total milk protein) (Flom et al. 2019, Villa et al. 2018).
Bos d 6 (type of whey protein) is present in minor quantities in CM (Villa et al. 2018). It (1.2% of whey content) penetrates the milk via secretory cells from the blood (Jaiswal et al. 2021) and helps in the transport, metabolism and distribution of substances (like fatty acids, drugs, and hormones). Furthermore, this protein also plays a role in regulating the colloidal osmotic pressure and protects from free radicals (Villa et al. 2018).
According to WHO/IUIS, Bos d 6 (serum albumin) is listed as a food allergen from CM (WHO/IUIS 2019).
Risk factors
Allery to Bos d 6 in patients with persistent CMA is considered a risk factor for developing asthma and rhinoconjunctivitis due to animal epithelia sensitization (Vicente-Serrano et al. 2007).
Clinical Relevance
Disease severity
CMA is considered as a major common pediatric food allergy (Agyemang et al. 2019). Milk allergy can be either IgE-mediated or non-IgE-mediated (Walsh et al. 2016). The clinical manifestations of CM protein allergy are reported to be of two types such as immediate (shortly after consumption) and delayed (after hours or days of consumption) responses (De Greef et al. 2012). The immediate reactions are generally IgE-mediated and can lead to angioedema, urticaria, aggravation of atopic dermatitis (AD), vomiting, in some cases, anaphylaxis, as well as respiratory symptoms (breathing, wheezing, coughing) (De Greef et al. 2012, Flom et al. 2019, Ito et al. 2012, Walsh et al. 2016). On the other hand, most delayed reactions are non-IgE mediated (Walsh et al. 2016) and can lead to gastrointestinal symptoms (like enteropathy or proctocolitis) or skin reactions (such as AD) (De Greef et al. 2012, Ito et al. 2012).
As per a study conducted in Iran, anaphylactic reactions were observed in 60% of the patients with CMA showing a positive test for sIgE towards Bos d 6 (Shokouhi Shoormasti et al. 2016).
Cross-reactive molecules
BSA is a minor allergen in milk (Bos d 6) but a major allergen in beef. Thus, patients with milk allergy sensitized to Bos d 6 may have a concomitant beef allergy, and BSA has also been found to cross-react with other BSA such as pork and sheep (Martelli et al. 2002, Vicente-Serrano et al. 2007, Werfel et al. 1997).
A prospective cohort study conducted on 38 children (age <14 years) with CMA-associated symptoms detected the rate of cross-sensitization (SPT-based) between goat milk and cow milk to be 63.2% (24/38) (Ehlayel et al. 2011).
According to a study, the cross-reactivity found between SA of cow, pork, deer, sheep, dog, and, cat (meat and/or epithelia) in the sera of all the patients with persistent milk allergy (n=8) was inhibited by pure Bos d 6 (Vicente-Serrano et al. 2007).
An Egypt-based study reported cross-reactivity between CM and beef in 30 patients with CMA (Refaat et al. 2011).
Molecular Aspects
Biochemistry
Bos d 6 (BSA) is a protein (molecular weight: 67 kDa) consisting of 583 amino acid residues and 35 Cys residues (Inge et al. 2022, WHO/IUIS 2019, Wynn et al. 2022). This protein is arranged into 3 homologous domains and comprises of 9 loops linked by 17 covalent disulfide bonds. Most of these disulfide bonds are protected inside the core of the protein, making them less accessible. Even under denaturing conditions, disulfide bonds help maintain the native antigenic determinants in BSA due to their stable tertiary structure (Farrell et al. 2004, Restani et al. 1999, Villa et al. 2018).
BSA is water-soluble and rich in amino acids recognized by gastrointestinal enzymes; however, it is comparatively resistant to digestion (Restani et al. 1999).
Isoforms, epitopes, antibodies
Bos d 6 has been reported to possess at least one sequential epitope, corresponding to amino acids 524-542 (crucial for antibody binding) (Chruszcz et al. 2013, Restani et al. 1999).
To date (25th August 2022), only one isoallergen of Bos d 6, i.e., Bos d 6.0101, has been identified and officially published by the WHO/IUIS (WHO/IUIS 2019).
Bos d 6 is the most heat-labile of all the milk proteins (El-Agamy 2007). In a study, 8 adults with a positive response to oral challenge with milk were subjected to boiled milk (10 minutes). Patients reactive to BSA exhibited no SPT response to boiled milk, thus showing reduced whey protein antigenicity (A Norgaard; H 1996, Restani et al. 2009).
Cross-reactivity due to structural similarity
Milk of different ruminant species (such as cow, goat, and sheep) is composed of homologous proteins with similar structures and functions, thereby leading to cross-reactivity between the milk proteins (Villa et al. 2018).
Bos d 6 from CM protein shows sequence homology with other mammalian milk, including ewe (92.4%), goat (71.2%), pig (79.9%), donkey (74.1%), horse (74.5%) and human (76.6%) (Tsabouri et al. 2014).
Diagnostic Relevance
Disease Severity
In an Italy-based study, 92.9% of children with beef allergy were also allergic to CM. Thus, sensitization to Bos d 6 was a predictive marker of CMA in children with beef allergy (Martelli et al. 2002).
Exposure
Cow milk allergy mainly occurs through ingestion (Kaczmarski et al. 2013).
Explained Results
Allergen information
Bos d 6 (bovine serum albumin) is a minor allergenic protein found in CM and is the most heat-labile protein of all the milk proteins (Villa et al. 2018).
Clinical relevance
Typical symptoms of CMA include skin reactions (angioedema, urticaria, and atopic dermatitis), GI symptoms, respiratory systems, and in some cases, anaphylaxis (De Greef et al. 2012, Flom et al. 2019, Ito et al. 2012, Shokouhi Shoormasti et al. 2016).
Cross-reactivity
Bos d 6 (BSA) from CM has been found to show cross-reactivity with SA of deer, pork, sheep, dog, and cat (meat and/or epithelia) (Vicente-Serrano et al. 2007). Moreover, cross-reactivity has also been reported between CM and beef (Refaat et al. 2011).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Eva Södergren
References
A Norgaard; H, B. "Allergenicity of individual cow milk proteins in DBPCFC-positive milk allergic adults." Journal of Allergy and Clinical Immunology   1996 97(1 Part 3): 237
Agyemang, A., et al. "Utilizing boiled milk sIgE as a predictor of baked milk tolerance in cow's milk allergic children." J Allergy Clin Immunol Pract   2019 7(6): 2049-2051.10.1016/j.jaip.2019.01.034.
Chruszcz, M., et al. "Serum albumins-unusual allergens." Biochim Biophys Acta   2013 1830(12): 5375-5381.10.1016/j.bbagen.2013.06.016.
De Greef, E., et al. "Diagnosis and management of cow’s milk protein allergy in infants." World Journal of Pediatrics   2012 8(1): 19-24
Ehlayel, M., et al. "Camel milk is a safer choice than goat milk for feeding children with cow milk allergy." ISRN Allergy   2011 2011: 391641.10.5402/2011/391641.
El-Agamy, E. "The challenge of cow milk protein allergy." Small Ruminant Research   2007 68(1-2): 64-72
Farrell, H. M., Jr., et al. "Nomenclature of the proteins of cows' milk--sixth revision." J Dairy Sci   2004 87(6): 1641-1674.10.3168/jds.S0022-0302(04)73319-6.
Flom, J. D. and S. H. Sicherer "Epidemiology of Cow's Milk Allergy." Nutrients   2019 11(5).10.3390/nu11051051.
Gjesing, B., et al. "Allergen-specific IgE antibodies against antigenic components in cow milk and milk substitutes." Allergy   1986 41(1): 51-56.10.1111/j.1398-9995.1986.tb00275.x.
Hochwallner, H., et al. "Microarray and allergenic activity assessment of milk allergens." Clin Exp Allergy   2010 40(12): 1809-1818.10.1111/j.1365-2222.2010.03602.x.
Hochwallner, H., et al. "Cow's milk allergy: from allergens to new forms of diagnosis, therapy and prevention." Methods   2014 66(1): 22-33.10.1016/j.ymeth.2013.08.005.
Host, A., et al. "Prospective estimation of IgG, IgG subclass and IgE antibodies to dietary proteins in infants with cow milk allergy. Levels of antibodies to whole milk protein, BLG and ovalbumin in relation to repeated milk challenge and clinical course of cow milk allergy." Allergy   1992 47(3): 218-229.10.1111/j.1398-9995.1992.tb00654.x.
Inge, G., et al. "Heterogeneity, Fractionation, and Isolation."   2022: 881-893.https://doi.org/10.1016/B978-0-12-818766-1.00278-6.
Ito, K., et al. "The usefulness of casein-specific IgE and IgG4 antibodies in cow's milk allergic children." Clin Mol Allergy   2012 10(1): 1.10.1186/1476-7961-10-1.
Jaiswal, L. and M. Worku "Recent perspective on cow's milk allergy and dairy nutrition." Crit Rev Food Sci Nutr   2021: 1-16.10.1080/10408398.2021.1915241.
Kaczmarski, M., et al. "The natural history of cow's milk allergy in north-eastern Poland." Adv Med Sci   2013 58(1): 22-30.10.2478/v10039-012-0053-3.
Martelli, A., et al. "Beef allergy in children with cow's milk allergy; cow's milk allergy in children with beef allergy." Ann Allergy Asthma Immunol   2002 89(6 Suppl 1): 38-43.10.1016/s1081-1206(10)62121-7.
Muthukumar, J., et al. "Food and food products associated with food allergy and food intolerance - An overview." Food Res Int   2020 138(Pt B): 109780.10.1016/j.foodres.2020.109780.
Natale, M., et al. "Cow's milk allergens identification by two-dimensional immunoblotting and mass spectrometry." Mol Nutr Food Res   2004 48(5): 363-369.10.1002/mnfr.200400011.
Refaat, M., et al. "Beef Meat Allergy in Cow's Milk Allergic Adults." Food and Nutrition Sciences   2011 02.10.4236/fns.2011.28121.
Restani, P., et al. "Molecular aspects of milk allergens and their role in clinical events." Anal Bioanal Chem   2009 395(1): 47-56.10.1007/s00216-009-2909-3.
Restani, P., et al. "Cross-reactivity between milk proteins from different animal species." Clin Exp Allergy   1999 29(7): 997-1004.10.1046/j.1365-2222.1999.00563.x.
Shokouhi Shoormasti, R., et al. "The Most Common Cow's Milk Allergenic Proteins with Respect to Allergic Symptoms in Iranian Patients." Iran J Allergy Asthma Immunol   2016 15(2): 161-165
Shoormasti, R. S., et al. "The most common cow’s milk allergenic proteins regarding to allergic symptoms." Clinical and Translational Allergy   2011 1(1): P62.10.1186/2045-7022-1-S1-P62.
Skripak, J. M., et al. "The natural history of IgE-mediated cow's milk allergy." J Allergy Clin Immunol   2007 120(5): 1172-1177.10.1016/j.jaci.2007.08.023.
Tsabouri, S., et al. "Cow's milk allergenicity." Endocr Metab Immune Disord Drug Targets   2014 14(1): 16-26.10.2174/1871530314666140121144224.
Vandenplas, Y. "Prevention and Management of Cow's Milk Allergy in Non-Exclusively Breastfed Infants." Nutrients   2017 9(7).10.3390/nu9070731.
Vicente-Serrano, J., et al. "Sensitization to serum albumins in children allergic to cow's milk and epithelia." Pediatr Allergy Immunol   2007 18(6): 503-507.10.1111/j.1399-3038.2007.00548.x.
Villa, C., et al. "Bovine Milk Allergens: A Comprehensive Review." Comprehensive Reviews in Food Science and Food Safety   2018 17(1): 137-164.https://doi.org/10.1111/1541-4337.12318.
Wal, J.-M. "Cow's milk proteins/allergens." Annals of Allergy, Asthma & Immunology   2002 89(6, Supplement): 3-10.https://doi.org/10.1016/S1081-1206(10)62115-1.
Walsh, J., et al. "Differentiating milk allergy (IgE and non-IgE mediated) from lactose intolerance: understanding the underlying mechanisms and presentations." Br J Gen Pract   2016 66(649): e609-611.10.3399/bjgp16X686521.
Werfel, S. J., et al. "Clinical reactivity to beef in children allergic to cow's milk." J Allergy Clin Immunol   1997 99(3): 293-300.10.1016/s0091-6749(97)70045-9.
WHO/IUIS. (2019). "Bos d 6."   Retrieved 09-06-2022, from http://www.allergen.org/viewallergen.php?aid=164.
Wynn, P. C. and P. A. Sheehy "Milk Proteins: Minor Proteins, Bovine Serum Albumin, and Vitamin-Binding Proteins and their Biological Properties☆."   2022: 942-947.https://doi.org/10.1016/B978-0-12-818766-1.00192-6.