+
For Healthcare Professionals
For Lab Professionals
+
For Patients & Caregivers
For Lab Professionals
+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Healthcare or Laboratory Professional content
Welcome! Click here for Patient or Laboratory Professional content
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Meet Antonio & Isabel
Antonio and Isabel, 18-month-olds, who both have a history of eczema and milk allergy visit their healthcare professional for additional insight into their milk allergy.
Antonio and Isabel's healthcare professional conducts a full clinical history and physical examination and decides to test using milk whole allergen with component reflex.
Patient History – Antonio
Family history
  • None 
Personal history 
  • Severe eczema since 2– 3 months of age
  • Stomach ache and loose stools 
Skin prick test results at 6 months
Test Type Results
Milk Whole Allergen +4
Negative skin prick test to egg
ImmunoCAPTM results (kUA /l) at 6 months
Test Type Results
Milk Whole Allergen 18
His previous physician recommended eliminating cow’s milk from the diet and substitute with milk-free formula
Patient History – Isabel
Family history
  • None 
Personal history 
  • Severe eczema since 2– 3 months of age
  • Stomach ache and loose stools 
Skin prick test results at 6 months
Test Type Results
Milk Whole Allergen +5
Negative skin prick test to egg
ImmunoCAP results (kUA /l) at 6 months
Test Type Results
Milk Whole Allergen 25
Her previous physician recommended eliminating cow’s milk from the diet and substitute with milk-free formula 
New Test results – Antonio
These results, together with this patient's case history and symptoms, help confirm the diagnosis.  
ImmunoCAP results (kUA /l)
Test Type Results
Milk Whole Allergen 10.2
Bos d 4 (α-lactalbumin) Allergen Component 3.8
Bos d 5 (β-lactoglobulin) Allergen Component 0.7
Bos d 8 (casein) Allergen Component 1.4
New Test results – Isabel
These results, together with this patient's case history and symptoms, help confirm the diagnosis. 
ImmunoCAP results (kUA /l)
Test Type Results
Milk Whole Allergen 20.6
Bos d 4 (α-lactalbumin) Allergen Component 2.2
Bos d 5 (β-lactoglobulin) Allergen Component 1.0
Bos d 8 (casein) Allergen Component 21.5
Differential diagnosis - Antonio
Antonio's low level of sensitization to Bos d 8 casein indicates he may have a tolerance to extensively heated milk. An oral challenge with extensively heated milk may be performed to confirm the diagnosis. With components and follow-up oral challenge, Antonio's quality of life could have been improved: No need of strict avoidance of milk and no cause of stress in the family due to the risk of eating products containing extensively heated milk, such as cakes and cookies.1,2
Refined diagnosis:
  • Likely tolerant to baked milk
Healthcare professional's treatment plan for Antonio
  • Antonio's healthcare professional advises him to undergo an oral food challenge to confirm tolerance to baked milk
Follow Up
Antonio is seen by his healthcare professional 1 year later—he had successfully passed the oral food challenge for milk and has been eating foods containing extensively heated milk since with no issues.
Differential diagnosis - Isabel
Isabel's significant levels of IgE to Bos d 8 indicates a risk for severe reaction to all forms of milk.1,2
Refined diagnosis:
  • Milk allergy
Healthcare Professional's management plan for Isabel
  • Dietary restriction to all forms of milk remains
Follow Up
Isabel is retested one year later, and her results are the same. She has continued to avoid milk in all forms and is doing well.
The people, places and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
Refrences
  1. Nowak-Wegrzyn A, Bloom KA, Sicherer SH, et al. Tolerance to extensively heated milk in children with cow’s milk allergy. J Allergy Clin Immunol. 2008;122(2):342-347 
  2. Tosca MA, Pistorio A, Rossi GA, Ciprandi G. Cow’s Milk Allergy: the Relevance of IgE. Allergy, Asthma & Immunology Research. 2016;8(1):86-87. doi:10.4168/aair.2016.8.1.86.