+
For Healthcare Professionals
For Lab Professionals
+
For Patients & Caregivers
For Lab Professionals
+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Healthcare or Laboratory Professional content
Welcome! Click here for Patient or Laboratory Professional content
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Meet Elizabeth
Elizabeth, a 5-year-old with a milk allergy, visits her healthcare professional for her annual check-up. Since 75% of children with cow’s milk allergy can tolerate baked foods containing milk,1,2 Elizabeth's healthcare professional suggested she should be tested with milk and to include milk component testing.
Patient History
Family history
  • None
Personal history
  • Diagnosed with eczema at her 6-month check up
  • Gastrointestinal distress, vomiting and eczema worsening after drinking milk at 3
  • Tested with pediatric allergy profile including foods and indoor respiratory allergens. Results confirmed sensitization to milk
Test Results
These results, together with this patient's case history and symptoms, help confirm the diagnosis.  
ImmunoCAPTM test results (kUA/l)
Test Type Results
Milk Whole Allergen 2.13
Bos d 4 (α-lactalbumin) Allergen Component 1.22
Bos d 5 (β-lactoglobulin) Allergen Component <.10 
Bos d 8 (casein) Allergen Component <.10
Differential Diagnosis
Elizabeth is sensitized to α-lactalbumin proteins. Her risk for reaction may be reduced when milk is thoroughly heated. She is not sensitized to casein proteins associated with severe reactions.1,2
Healthcare professional's management plan
  • Elizabeth's healthcare professional refers her to an allergy specialist for an oral food challenge with baked milk
  • The allergy specialist performs an oral baked milk challenge in the clinic, which Elizabeth passes without any symptoms.
  • The allergy specialist recommends that Elizabeth undergo retesting in about two years
Follow Up
Baked milk is now part of Elizabeth’s diet and she is doing well.
The people, places and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
References
  1. Nowak-Wegrzyn A, et al. Tolerance to extensively heated milk in children with cow's milk allergy. J Allergy Clin Immunol. 2008;122:342-7.
  2. Caubet JC, et al. Utility of casein-specific IgE levels in predicting reactivity to baked milk. J Allergy Clin Immunol. 2013;131:222-4.