+
For Healthcare Professionals
For Lab Professionals
+
For Patients & Caregivers
For Lab Professionals
+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Healthcare or Laboratory Professional content
Welcome! Click here for Patient or Laboratory Professional content
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Meet Jill
Jill, a 5-year old girl with a history of egg allergy, visits her healthcare professional after recently eating cake at a birthday party. Her mother reported that Jill experienced no apparent reaction after this event and is wondering if Jill has grown out of her egg allergy.
Jill's healthcare professional conducts a full clinical history and physical examination and decides to test using egg white with reflex to components.
Patient History
Family history
  • None
Personal history
  • Mild eczema as an infant after eating egg
  • Parents removed eggs from her diet at that point
Jill’s previous healthcare professional advised that Jill have testing done, but her parents didn’t want it at the time.
Test Results 
These results, together with this patient's case history and symptoms, help confirm the diagnosis.  
ImmunoCAPTM test results (kUA /l)
Test Type Results
Egg White Whole Allergen 6.4
Gal d 1 (Ovomucoid) Allergen Component <.10 
Gal d 2 (Ovalbumin) Allergen Component 5.42
Differential Diagnosis
Jill’s lack of IgE sensitization to Gal d 1 (ovomucoid) indicates that she may be able to eat extensively baked egg.1
An open challenge with baked egg (i.e. muffin) may be indicated.
Healthcare professional's management plan
  • Jill's healthcare professional refers her to an allergy specialist for an oral food challenge with baked egg
  • The allergy specialist performs a baked egg challenge in the clinic, which Jill passes without any symptoms.
  • Jill’s mother gradually re-introduces extensively baked egg, such as muffins or birthday cake, into her diet.
Follow Up
Jill with her mother are seen by her healthcare professional one year later. Extensively cooked egg is now part of Jill’s diet and she is fit and well. As she ages, Jill is likely to outgrow her egg allergy. Periodic testing may be appropriate to determine IF and WHEN Jill may outgrow her egg allergy all together. Depending on her sIgE results the allergist may determine when a baked egg challenge (i.e. scrambled eggs) may be indicated. Jill's mother is advised to use precaution when introducing certain foods that may contain raw egg, such as ice cream, salad dressings, and sauces, into her diet.
The people, places and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
References
  1. Montesinos E, Martorell A, Félix R, Cerdá JC. Egg white specific IgE levels in serum as clinical reactivity predictors in the course of egg allergy follow-up. Pediatr Allergy Immunol 2010: 21: 634–639.