Guidelines advise that diagnosis of food allergy starts with a physical examination and a food allergy-focused patient history1,2 in combination with diagnostic testing.3-5 An allergy-focused clinical history should be tailored to the presenting symptoms and age of the patient.2
Food allergy can be classified into IgE-mediated and non-IgE-mediated allergy. IgE-mediated reactions are acute and frequently have a rapid onset while non-IgE-mediated reactions are generally characterized by delayed and non-acute reactions.6
Guided by the allergy-focused patient history, work through the most appropriate next steps, including considerations for specific IgE tests—when added to an allergy-focused patient history, the use of skin-prick tests (SPT), serological tests, or both increases your confidence in diagnosis and clinical management by ruling in or ruling out allergies7,8
Key Allergy Test Differences | Specific IgE blood testing | Skin prick testing |
Typically ordered and reviewed by a clinician | YES | YES |
Patients do not need to discontinue allergy medications | YES | NO |
Requires only one needle stick (a single blood sample) | YES | NO |
Carries no risk of severe allergic reaction | YES | NO |
Can be used when extensive skin rash is present | YES | NO |
Can be used for children as young as 3 months of age | YES | YES |
Component testing is available |
YES | NO |
Same day results in office | NO | YES |
Eight allergens account for 90% of food allergies in children/young people.10,11
Milk
Wheat
Peanuts
Shellfish
Fish
Soy
Tree Nuts
Egg
If an allergy-focused clinical history suggests an IgE-mediated food allergy, SPT or specific IgE blood tests are needed to help confirm the diagnosis. A positive test on its own simply shows sensitization to a food allergen and is not itself diagnostic of food allergy.
Correct diagnosis of food allergy, followed by education and advice based on valid test results may help to reduce the incidence of adverse reactions resulting from true food allergies, and may also help to reduce the unnecessary dietary exclusion of foods that are safe and should be eaten as part of a normal, healthy diet.2
Patient presents with symptoms suggesting a food allergy
Allergy-focused patient history is taken
Allergy test for suspected
allergens
Only 1 ml of blood is required for the
main food allergen
Allergy management plan
Adapted from NICE Guideline CG1162
Consider an exclusion/re-introduction diet
consultation with a dietician
Consider an exclusion/re-introduction,
diagnosis is confirmed
Allergy management plan
Adapted from NICE Guideline CG1162
ImmunoCAPTM Whole Allergen testing provides an objective measurement of the circulating specific IgE antibodies and the sensitization to a specific whole allergen. Specific IgE antibodies can appear as a result of exposure to an allergen in atopic individuals. ImmunoCAP Whole Allergen measure IgE antibodies to specific allergens in human serum or plasma and allows quantitative measurements of a wide range of individual allergens. Over 550 different allergens are available for determinations.11
Often, allergies are not as clearcut as a simple “yes” or “no.” That’s where specific IgE blood testing for allergen components comes in. ImmunoCAPTM Allergen Components measure specific IgE antibodies to individual molecular allergens in serum or plasma. These allergens, which are purified or recombinant proteins, offer the unique opportunity to assess a person’s allergic sensitization pattern at the molecular level. Different component groups often elicit different types of reactions, so testing them can indicate if a patient’s positive test results were caused by allergy to a given substance or by a cross-reaction with another allergen. Specifically for peanuts and tree nuts; measuring specific IgE for allergen components helps the clinician weigh a patients risk of a systemic reaction versus a more mild or localized response.12
More than 100 different ImmunoCAP allergen components are available for determinations,11 including:
MILK
EGG
PEANUT
TREE NUTS
The presence of allergen-specific IgE is usually a risk of allergy symptoms and a result ≥0.1 kUA/L indicates sensitization. Traditionally, the higher the IgE level the greater the risk.
The higher the concentration of lgE antibodies, the higher the risk for symptomatic allergy16
Chart reference 15
Guided by the IgE-mediated test result, an individual plan to manage and treat the allergy can be tailored in conjunction with a dietitian, in order to achieve improved quality of life and overall well-being.
Results should be read in conjunction with the clinical history.13
The result of IgE-mediated tests can help to confirm a suspicion of allergy, determine the offending allergens, or to rule out allergy altogether. Ruling out allergy can be as important as confirming it. If allergy is ruled out as the cause of the symptoms, you may reduce worry and unnecessary food avoidance or medication.
You can also continue to search for other causes. Test results provide precise and reliable answers to a patient’s allergy profile which may help you discover hidden risks, such as allergies due to cross-reactions.